Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SIPA1L1 SIRNA Assays ›
          Gene Symbol:
          SIPA1L1
          Gene Name:
          signal induced proliferation associated 1 like 1
          Gene Aliases:
          E6TP1
          Chromosome Location:
          Chr.14: 71320418 - 71741229 on Build GRCh38
          UniGene ID:
          Hs.654657
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 23173

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SIPA1L1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001284245.1 NP_001271174.1 2, 3 1866
          NM_001284246.1 NP_001271175.1 2, 3 2455
          NM_001284247.1 NP_001271176.1 1, 2 1807
          NM_015556.2 NP_056371.1 2, 3 1866
          XM_005267516.4 XP_005267573.1 2, 3 2455
          XM_006720111.2 XP_006720174.1 4, 5 3745
          XM_011536630.1 XP_011534932.1 6, 7 2217
          XM_011536631.1 XP_011534933.1 6, 7 2628
          XM_011536632.1 XP_011534934.1 5, 6 2137
          XM_011536635.2 XP_011534937.1 5, 6 2284
          XM_017021159.1 XP_016876648.1 8, 9 2374
          XM_017021160.1 XP_016876649.1 6, 7 2296
          XM_017021161.1 XP_016876650.1 7, 8 2249
          XM_017021162.1 XP_016876651.1 9, 10 2431
          XM_017021163.1 XP_016876652.1 4, 5 2098
          XM_017021164.1 XP_016876653.1 4, 5 2105
          XM_017021165.1 XP_016876654.1 6, 7 2174
          XM_017021166.1 XP_016876655.1 7, 8 2307
          XM_017021167.1 XP_016876656.1 6, 7 2182
          XM_017021168.1 XP_016876657.1 6, 7 2156
          XM_017021169.1 XP_016876658.1 5, 6 2309
          XM_017021170.1 XP_016876659.1 5, 6 2667
          XM_017021171.1 XP_016876660.1 6, 7 2240
          XM_017021172.1 XP_016876661.1 5, 6 2115
          XM_017021173.1 XP_016876662.1 4, 5 2242
          XM_017021174.1 XP_016876663.1 4, 5 2482
          XM_017021175.1 XP_016876664.1 4, 5 2600
          XM_017021176.1 XP_016876665.1 9, 10 2427
          XM_017021177.1 XP_016876666.1 9, 10 2416
          XM_017021178.1 XP_016876667.1 7, 8 2254
          XM_017021179.1 XP_016876668.1 6, 7 2157
          XM_017021180.1 XP_016876669.1 10, 11 2469
          XM_017021181.1 XP_016876670.1 8, 9 2374
          XM_017021182.1 XP_016876671.1 7, 8 2307
          XM_017021183.1 XP_016876672.1 5, 6 2115
          XM_017021184.1 XP_016876673.1 9, 10 2427
          XM_017021185.1 XP_016876674.1 2, 3 2455
          XM_017021186.1 XP_016876675.1 8, 9 2374
          XM_017021187.1 XP_016876676.1 6, 7 2240
          XM_017021188.1 XP_016876677.1 5, 6 2115
          XM_017021189.1 XP_016876678.1 7, 8 2224
          XM_017021190.1 XP_016876679.1 8, 9 2374
          XM_017021191.1 XP_016876680.1 6, 7 2182
          XM_017021192.1 XP_016876681.1 5, 6 2115
          XM_017021193.1 XP_016876682.1 5, 6 2113
          XM_017021194.1 XP_016876683.1 8, 9 2374
          XM_017021195.1 XP_016876684.1 8, 9 2374
          XM_017021196.1 XP_016876685.1 8, 9 2374
          GenBank mRNA AB007900.2 - 1, 2 1590
          AF090989.1 - 2, 3 1853
          AF090990.1 - 2, 3 1853
          AK122930.1 - 2, 3 2455
          BC146777.1 - 1, 2 1590 9007226

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_139330, AY043226, AF026504.
          This siRNA also targets the following transcripts in Mouse: NM_001167983, NM_172579, AK122284, BC058681, AK051853, and others.

          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          actin cytoskeleton reorganization
          regulation of GTPase activity
          ephrin receptor signaling pathway
          regulation of synaptic plasticity
          regulation of axonogenesis
          regulation of small GTPase mediated signal transduction
          regulation of dendritic spine morphogenesis
          activation of GTPase activity
          molecular_function
          GTPase activator activity
          ephrin receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0