Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PRKAG2 SIRNA Assays ›
          Gene Symbol:
          PRKAG2
          Gene Name:
          protein kinase AMP-activated non-catalytic subunit gamma 2
          Gene Aliases:
          AAKG, AAKG2, CMH6, H91620p, WPWS
          Chromosome Location:
          Chr.7: 151556114 - 151877231 on Build GRCh38
          UniGene ID:
          Hs.647072
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 24040

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PRKAG2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001040633.1 NP_001035723.1 5, 6 1097
          NM_001304531.1 NP_001291460.1 2, 3 119
          NM_016203.3 NP_057287.2 5, 6 1366
          NM_024429.1 NP_077747.1 1, 2 191
          XM_005250002.3 XP_005250059.1 5, 6 1232
          XM_005250004.3 XP_005250061.1 5, 6 1178
          XM_005250006.4 XP_005250063.1 3, 4 931
          XM_006716021.2 XP_006716084.1 5, 6 769
          XM_011516283.1 XP_011514585.1 5, 6 769
          XM_011516285.1 XP_011514587.1 2, 3 611
          XM_017012270.1 XP_016867759.1 5, 6 756
          XM_017012273.1 XP_016867762.1 3, 4 916
          GenBank mRNA AB025580.1 - 1, 2 104
          AF087875.1 - 1, 2 152
          AJ249976.1 - 5, 6 845
          AK001887.1 - 1, 2 191
          AK308116.1 - 5, 6 797
          AK314646.1 - 1, 2 153
          AW664063.1 - 2, 3 91
          BC020540.1 - 1, 2 72
          BC068598.1 - 5, 6 1066 5259756
          BT007127.1 - 1, 2 32
          DA151645.1 - 2, 3 120

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          kinase modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          glycogen metabolic process
          regulation of glycolytic process
          protein phosphorylation
          negative regulation of protein kinase activity
          fatty acid biosynthetic process
          ATP biosynthetic process
          carnitine shuttle
          cell cycle arrest
          positive regulation of peptidyl-threonine phosphorylation
          sterol biosynthetic process
          macroautophagy
          regulation of fatty acid metabolic process
          activation of protein kinase activity
          intracellular signal transduction
          regulation of fatty acid biosynthetic process
          positive regulation of protein kinase activity
          regulation of fatty acid oxidation
          regulation of glucose import
          negative regulation of protein serine/threonine kinase activity
          regulation of signal transduction by p53 class mediator
          AMP-activated protein kinase activity
          cAMP-dependent protein kinase inhibitor activity
          ATP binding
          cAMP-dependent protein kinase regulator activity
          phosphorylase kinase regulator activity
          AMP binding
          protein kinase binding
          protein kinase activator activity
          ADP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0