Search
Search

SRRM2| Gene Symbol: SRRM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016333.3 | NP_057417.3 | 2 | 779 | ||
| XM_005255226.1 | XP_005255283.1 | 2 | 757 | |||
| XM_006720874.3 | XP_006720937.1 | 2 | 757 | |||
| GenBank mRNA | AB002322.3 | - | 2 | 431 | ||
| AB016089.1 | - | 2 | 415 | |||
| AB016092.1 | - | 2 | 455 | |||
| BC007752.2 | - | 2 | 431 | |||
| BC020639.1 | - | 2 | 470 | |||
| BC046104.1 | - | 2 | 435 | |||
| BC070050.1 | - | 2 | 490 | |||
| BX647088.1 | - | 1 | 325 | |||