Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CHD1L SIRNA Assays ›
          Gene Symbol:
          CHD1L
          Gene Name:
          chromodomain helicase DNA binding protein 1 like
          Gene Aliases:
          ALC1, CHDL
          Chromosome Location:
          Chr.1: 147172804 - 147295766 on Build GRCh38
          UniGene ID:
          Hs.191164
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 241441

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CHD1L
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256336.1 NP_001243265.1 3 386
          NM_004284.4 NP_004275.4 4 493
          NM_024568.2 NP_078844.2 4 465
          NR_046070.1 4 493
          XM_006711639.3 XP_006711702.1 3 361
          XM_017002858.1 XP_016858347.1 5 1221
          XM_017002859.1 XP_016858348.1 4 348
          XM_017002860.1 XP_016858349.1 4 441
          XM_017002861.1 XP_016858350.1 4 441
          XM_017002863.1 XP_016858352.1 3 256
          XM_017002864.1 XP_016858353.1 4 441
          GenBank mRNA AF054177.1 - 4 414
          AF537213.1 - 4 450
          AK001342.1 - 4 450
          AK027631.1 - 4 422
          AK223496.1 - 2 144
          AK298424.1 - 3 326
          AK307400.1 - 4 450
          BC001171.1 - 4 427
          BC005038.1 - 4 390
          BC043501.1 - 4 411
          CN255649.1 - 1 66
          DA007597.1 - 4 471
          DA155685.1 - 4 494
          DA745071.1 - 4 444
          EF560738.1 - 4 452
          KJ534809.1 - 4 430

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          nucleotide-excision repair, DNA duplex unwinding
          DNA repair
          nucleotide-excision repair, preincision complex stabilization
          nucleotide-excision repair, preincision complex assembly
          nucleotide-excision repair, DNA incision, 3'-to lesion
          nucleotide-excision repair, DNA incision, 5'-to lesion
          chromatin remodeling
          cellular response to DNA damage stimulus
          nucleotide-excision repair, DNA incision
          global genome nucleotide-excision repair
          nucleotide binding
          nucleic acid binding
          ATP-dependent DNA helicase activity
          protein binding
          ATP binding
          ATPase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0