Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPIP5K1 SIRNA Assays ›
Gene Symbol:
PPIP5K1
Gene Name:
diphosphoinositol pentakisphosphate kinase 1
Gene Aliases:
HISPPD2A, IP6K, IPS1, VIP1, hsVIP1
Chromosome Location:
Chr.15: 43533462 - 43590272 on Build GRCh38
UniGene ID:
Hs.156814
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
241546
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPIP5K1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130858.2
NP_001124330.1
6, 7
731
NM_001130859.2
NP_001124331.1
5, 6
726
NM_001190214.1
NP_001177143.1
6, 7
731
NM_014659.5
NP_055474.3
5, 6
649
XM_005254803.1
XP_005254860.1
6, 7
731
XM_005254804.1
XP_005254861.1
6, 7
731
XM_006720786.2
XP_006720849.1
6, 7
731
XM_011522249.1
XP_011520551.1
6, 7
731
XM_011522250.1
XP_011520552.1
6, 7
834
XM_011522252.1
XP_011520554.1
5, 6
705
XM_011522254.1
XP_011520556.1
5, 6
601
XM_011522259.1
XP_011520561.1
6, 7
731
XM_017022735.1
XP_016878224.1
5, 6
1048
XM_017022736.1
XP_016878225.1
5, 6
1016
XM_017022737.1
XP_016878226.1
5, 6
697
XM_017022738.1
XP_016878227.1
5, 6
645
XM_017022739.1
XP_016878228.1
5, 6
645
XM_017022740.1
XP_016878229.1
6, 7
834
XM_017022741.1
XP_016878230.1
5, 6
1047
XM_017022742.1
XP_016878231.1
6, 7
839
XM_017022743.1
XP_016878232.1
5, 6
705
XM_017022744.1
XP_016878233.1
5, 6
646
XM_017022745.1
XP_016878234.1
6, 7
834
XM_017022746.1
XP_016878235.1
5, 6
705
XM_017022747.1
XP_016878236.1
5, 6
645
XM_017022748.1
XP_016878237.1
5, 6
1047
XM_017022749.1
XP_016878238.1
6, 7
807
XM_017022750.1
XP_016878239.1
5, 6
648
XM_017022751.1
XP_016878240.1
5, 6
601
XM_017022752.1
XP_016878241.1
6, 7
731
XM_017022753.1
XP_016878242.1
5, 6
705
XM_017022754.1
XP_016878243.1
6, 7
839
XM_017022755.1
XP_016878244.1
5, 6
705
XM_017022756.1
XP_016878245.1
5, 6
643
XM_017022757.1
XP_016878246.1
5, 6
601
XM_017022759.1
XP_016878248.1
5, 6
645
XM_017022760.1
XP_016878249.1
5, 6
664
GenBank mRNA
AB002375.1
-
5, 6
653
AF502586.1
-
5, 6
621
AF502587.1
-
5, 6
653
AF502588.1
-
5, 6
621
AF502589.1
-
5, 6
653
AF543190.1
-
5, 6
603
AK307438.1
-
6, 7
695
BC050263.1
-
4, 5
677
5521199
BC057395.1
-
5, 6
677
6142663
BX538022.1
-
5, 6
677
BX647814.1
-
6, 7
686
Back To Top
More Information
Panther Classification:
Molecular Function -
nucleotide kinase
Gene Ontology Categories:
Function(s)
Process(es)
inositol metabolic process
phosphorylation
dephosphorylation
inositol phosphate biosynthetic process
inositol phosphate metabolic process
inositol-1,3,4,5,6-pentakisphosphate kinase activity
inositol hexakisphosphate kinase activity
inositol heptakisphosphate kinase activity
inositol hexakisphosphate 5-kinase activity
acid phosphatase activity
ATP binding
diphosphoinositol-pentakisphosphate kinase activity
inositol hexakisphosphate 1-kinase activity
inositol hexakisphosphate 3-kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0