Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DPY19L2 SIRNA Assays ›
Gene Symbol:
DPY19L2
DPY19L2P1
Gene Name:
dpy-19 like 2
DPY19L2 pseudogene 1
Gene Aliases:
SPATA34, SPGF9
-
Chromosome Location:
Chr.12: 63558913 - 63669201 on Build GRCh38
Chromosome Location:
Chr.7: 35081287 - 35186162 on Build GRCh38
UniGene ID:
Hs.533644
Hs.594815
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
242048
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
286.00
Your Price
Online offer:
277.65
277.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DPY19L2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_173812.4
NP_776173.3
22
2639
XM_006719352.2
XP_006719415.1
22
2565
XM_011538215.2
XP_011536517.1
20
2105
XM_011538218.2
XP_011536520.1
15
1860
XM_017019188.1
XP_016874677.1
22
2686
XM_017019189.1
XP_016874678.1
21
2626
XM_017019190.1
XP_016874679.1
21
2560
XM_017019191.1
XP_016874680.1
20
2536
XM_017019192.1
XP_016874681.1
20
2524
XM_017019193.1
XP_016874682.1
21
2221
XM_017019194.1
XP_016874683.1
21
2248
XM_017019195.1
XP_016874684.1
22
2296
XM_017019196.1
XP_016874685.1
20
2385
XM_017019197.1
XP_016874686.1
21
2519
XM_017019198.1
XP_016874687.1
18
2240
XM_017019199.1
XP_016874688.1
18
2309
XM_017019201.1
XP_016874690.1
21
2482
XM_017019202.1
XP_016874691.1
20
2095
XM_017019203.1
XP_016874692.1
20
2317
XM_017019204.1
XP_016874693.1
21
2555
XM_017019205.1
XP_016874694.1
18
2409
GenBank mRNA
AK057511.1
-
22
2610
AK303727.1
-
6
1100
AY358792.1
-
22
2560
BC031225.1
-
16
1641
BC068442.1
-
22
2642
5297524
Gene Symbol: DPY19L2P1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_002833.2
22
2692
GenBank mRNA
BC066987.1
-
22
2692
BX648666.1
-
18
1927
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
multicellular organism development
spermatid development
protein C-linked glycosylation via 2'-alpha-mannosyl-L-tryptophan
mannosyltransferase activity
transferase activity, transferring glycosyl groups
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0