Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CCDC88A SIRNA Assays ›
          Gene Symbol:
          CCDC88A
          Gene Name:
          coiled-coil domain containing 88A
          Gene Aliases:
          APE, GIRDIN, GIV, GRDN, HkRP1, KIAA1212
          Chromosome Location:
          Chr.2: 55287817 - 55419921 on Build GRCh38
          UniGene ID:
          Hs.292925
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 24919

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CCDC88A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001135597.1 NP_001129069.1 5 1199
          NM_001254943.1 NP_001241872.1 5 1199
          NM_018084.4 NP_060554.3 5 1199
          XM_005264418.4 XP_005264475.1 5 902
          XM_005264421.2 XP_005264478.1 5 902
          XM_005264426.2 XP_005264483.1 5 902
          XM_011532965.2 XP_011531267.1 5 902
          XM_011532966.2 XP_011531268.1 5 902
          XM_011532967.2 XP_011531269.1 5 902
          XM_011532968.2 XP_011531270.1 5 902
          XM_017004476.1 XP_016859965.1 5 902
          XM_017004477.1 XP_016859966.1 5 902
          XM_017004478.1 XP_016859967.1 5 902
          XM_017004479.1 XP_016859968.1 5 902
          XM_017004480.1 XP_016859969.1 5 902
          XM_017004481.1 XP_016859970.1 5 902
          GenBank mRNA AB125644.1 - 5 357
          AB201172.1 - 5 891
          AF112218.1 - 5 890
          AK024717.1 - 3 104
          AK025723.1 - 5 1133
          AK307964.1 - 5 597
          BC041160.1 - 5 1105
          BC060037.1 - 5 736
          BC078168.1 - 5 349
          BC132736.1 - 5 554 40146758
          BC144320.1 - 5 554 9052840
          D64159.1 - 5 890

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of protein phosphorylation
          DNA replication
          regulation of DNA replication
          regulation of neuron projection development
          cell migration
          lamellipodium assembly
          TOR signaling
          activation of protein kinase B activity
          regulation of actin cytoskeleton organization
          regulation of cell proliferation
          membrane organization
          actin binding
          microtubule binding
          phosphatidylinositol binding
          protein homodimerization activity
          protein kinase B binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0