Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other POU6F1 SIRNA Assays ›
          Gene Symbol:
          POU6F1
          Gene Name:
          POU class 6 homeobox 1
          Gene Aliases:
          BRN5, MPOU, TCFB1
          Chromosome Location:
          Chr.12: 51186936 - 51218134 on Build GRCh38
          UniGene ID:
          Hs.555886
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 251310

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: POU6F1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002702.3 NP_002693.3 6 1297
          NR_026893.1 11 1755
          XM_017019507.1 XP_016874996.1 11 2942
          XM_017019508.1 XP_016874997.1 12 2853
          XM_017019509.1 XP_016874998.1 10 1862
          XM_017019510.1 XP_016874999.1 11 2291
          XM_017019511.1 XP_016875000.1 11 2460
          XM_017019512.1 XP_016875001.1 11 1816
          XM_017019513.1 XP_016875002.1 11 2939
          XM_017019514.1 XP_016875003.1 11 2936
          XM_017019515.1 XP_016875004.1 11 2282
          XM_017019516.1 XP_016875005.1 11 2933
          XM_017019517.1 XP_016875006.1 10 2813
          XM_017019518.1 XP_016875007.1 10 2807
          XM_017019519.1 XP_016875008.1 10 2767
          XM_017019520.1 XP_016875009.1 9 2169
          XM_017019521.1 XP_016875010.1 9 2188
          XM_017019522.1 XP_016875011.1 10 1666
          XM_017019523.1 XP_016875012.1 8 1373
          XM_017019524.1 XP_016875013.1 10 1764
          XM_017019525.1 XP_016875014.1 11 1752
          XM_017019526.1 XP_016875015.1 10 1660
          XM_017019527.1 XP_016875016.1 10 1679
          XM_017019528.1 XP_016875017.1 11 2185
          XM_017019529.1 XP_016875018.1 10 2071
          XM_017019530.1 XP_016875019.1 9 1976
          GenBank mRNA AK095191.1 - 11 1755
          AL832881.1 - 6 1297
          BC051326.1 - 6 1100 6275848
          BC074765.2 - 5 840 30915178
          BC107090.1 - 1 172
          L14482.1 - 6 968
          Z21966.1 - 5 1007

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          brain development
          heart development
          muscle organ development
          RNA polymerase II regulatory region sequence-specific DNA binding
          transcription factor activity, sequence-specific DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-zxk44:80/100.66.78.82:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0