Search
Search

Dap3| Gene Symbol: Dap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011950.2 | NP_001011950.2 | 13 | 1648 | ||
| XM_006232610.2 | XP_006232672.1 | 12 | 1354 | |||
| GenBank mRNA | BC085878.1 | - | 13 | 1648 | 7191747 | |
| FQ230674.1 | - | 13 | 1437 | |||