Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PAK3 SIRNA Assays ›
Gene Symbol:
PAK3
Gene Name:
p21 (RAC1) activated kinase 3
Gene Aliases:
ARA, MRX30, MRX47, OPHN3, PAK-3, PAK3beta, bPAK, beta-PAK
Chromosome Location:
Chr.X: 110944241 - 111227361 on Build GRCh38
UniGene ID:
Hs.593599
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
255
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PAK3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128166.2
NP_001121638.1
10
1334
NM_001128167.2
NP_001121639.1
9
1261
NM_001128168.2
NP_001121640.1
14
1583
NM_001128172.2
NP_001121644.1
9
936
NM_001128173.2
NP_001121645.1
13
1520
NM_001324325.1
NP_001311254.1
10
1412
NM_001324326.1
NP_001311255.1
10
1257
NM_001324327.1
NP_001311256.1
12
1369
NM_001324328.1
NP_001311257.1
11
1302
NM_001324329.1
NP_001311258.1
13
1517
NM_001324330.1
NP_001311259.1
10
1409
NM_001324331.1
NP_001311260.1
11
1324
NM_001324332.1
NP_001311261.1
12
1475
NM_001324333.1
NP_001311262.1
12
1450
NM_001324334.1
NP_001311263.1
11
1405
NM_002578.4
NP_002569.1
12
1472
NR_136747.1
11
1324
NR_136748.1
9
1021
XM_005262132.2
XP_005262189.1
13
1512
XM_005262133.3
XP_005262190.1
12
1467
XM_006724654.2
XP_006724717.1
12
1359
XM_006724655.2
XP_006724718.1
11
1302
XM_011530962.1
XP_011529264.1
13
1510
XM_011530967.2
XP_011529269.1
12
1450
XM_011530968.2
XP_011529270.1
13
1526
XM_011530969.1
XP_011529271.1
11
1371
XM_017029557.1
XP_016885046.1
11
1371
XM_017029558.1
XP_016885047.1
11
1314
XM_017029559.1
XP_016885048.1
10
1326
XM_017029560.1
XP_016885049.1
10
1257
XM_017029561.1
XP_016885050.1
12
1474
XM_017029562.1
XP_016885051.1
13
1512
XM_017029563.1
XP_016885052.1
14
1575
GenBank mRNA
AB102659.1
-
9
891
AF068864.1
-
8
846
AF155651.1
-
12
1237
AK128022.1
-
11
1579
AK290504.1
-
9
1261
AK315284.1
-
10
1178
AM943850.1
-
9
891
AM943851.1
-
9
909
AM943852.1
-
10
954
BC117353.1
-
9
940
BC117355.1
-
9
940
BC143544.1
-
9
940
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPK activity
mitotic cell cycle
stimulatory C-type lectin receptor signaling pathway
axonogenesis
dendrite development
regulation of actin filament polymerization
T cell costimulation
positive regulation of neuron apoptotic process
ephrin receptor signaling pathway
synapse organization
T cell receptor signaling pathway
dendritic spine morphogenesis
positive regulation of dendritic spine morphogenesis
cellular response to organic cyclic compound
positive regulation of DNA biosynthetic process
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
MAP kinase kinase activity
protein binding
ATP binding
Rho GTPase binding
SH3 domain binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0