Search
Search

LOC100910882
Rbm39| Gene Symbol: LOC100910882 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749605.3 | XP_003749653.1 | 16, 17 | 1810 | ||
| XM_003749606.3 | XP_003749654.1 | 16, 17 | 1792 | |||
| XM_006235443.2 | XP_006235505.1 | 16, 17 | 1831 | |||
| XM_006235446.2 | XP_006235508.1 | 16, 17 | 3252 | |||
| XM_008762415.1 | XP_008760637.1 | 15, 16 | 1747 | |||
| XM_008762416.1 | XP_008760638.1 | 15, 16 | 1768 | |||
| XM_008762417.1 | XP_008760639.1 | 15, 16 | 7650 | |||
| XM_008762418.1 | XP_008760640.1 | 15, 16 | 4742 | |||
| XM_008762419.1 | XP_008760641.1 | 15, 16 | 6384 | |||
| XM_008762420.1 | XP_008760642.1 | 15, 16 | 7671 | |||
| XM_008762421.1 | XP_008760643.1 | 16, 17 | 6160 | |||
| XM_008762422.1 | XP_008760644.1 | 15, 16 | 1729 | |||
| Gene Symbol: Rbm39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013207.2 | NP_001013225.1 | 1734 | |||
| XM_006235331.2 | XP_006235393.1 | 1810 | ||||
| XM_006235332.2 | XP_006235394.1 | 1831 | ||||
| XM_006235335.2 | XP_006235397.1 | 3252 | ||||
| XM_008762349.1 | XP_008760571.1 | 1747 | ||||
| XM_008762350.1 | XP_008760572.1 | 1768 | ||||
| XM_008762351.1 | XP_008760573.1 | 15, 16 | 7650 | |||
| XM_008762352.1 | XP_008760574.1 | 15, 16 | 4742 | |||
| XM_008762353.1 | XP_008760575.1 | 6384 | ||||
| XM_008762354.1 | XP_008760576.1 | 15, 16 | 7671 | |||
| XM_008762355.1 | XP_008760577.1 | 6160 | ||||
| XM_008762356.1 | XP_008760578.1 | 1729 | ||||
| GenBank mRNA | AY769432.1 | - | 604 | |||
| BC078917.1 | - | 1825 | ||||
| BC091394.1 | - | 1723 | 7322533 | |||