Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SYBU SIRNA Assays ›
Gene Symbol:
SYBU
Gene Name:
syntabulin
Gene Aliases:
GOLSYN, OCSYN, SNPHL
Chromosome Location:
Chr.8: 109574176 - 109691791 on Build GRCh38
UniGene ID:
Hs.390738
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
25640
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SYBU
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001099743.1
NP_001093213.1
4, 5
948
NM_001099744.1
NP_001093214.1
4, 5
951
NM_001099745.1
NP_001093215.1
4, 5
787
NM_001099746.1
NP_001093216.1
2, 3
501
NM_001099747.1
NP_001093217.1
4, 5
705
NM_001099748.1
NP_001093218.1
4, 5
708
NM_001099749.1
NP_001093219.1
2, 3
422
NM_001099750.1
NP_001093220.1
5, 6
829
NM_001099751.1
NP_001093221.1
4, 5
794
NM_001099752.1
NP_001093222.1
4, 5
797
NM_001099753.1
NP_001093223.1
3, 4
595
NM_001099754.1
NP_001093224.1
3, 4
598
NM_001099755.1
NP_001093225.1
2, 3
393
NM_001099756.1
NP_001093226.1
2, 3
669
NM_017786.5
NP_060256.3
5, 6
826
XM_017013614.1
XP_016869103.1
3, 4
517
XM_017013615.1
XP_016869104.1
3, 4
491
GenBank mRNA
AB040905.1
-
2, 3
251
AB195676.1
-
5, 6
759
AB195677.1
-
4, 5
638
AB195678.1
-
2, 3
355
AB195679.1
-
2, 3
1154
AK000373.1
-
2, 3
480
AK055246.1
-
4, 5
718
AK090559.1
-
2, 3
388
AK096558.1
-
3, 4
593
AK290469.1
-
4, 5
787
AL713645.1
-
5, 6
720
AY705385.1
-
3, 4
435
BC012082.1
-
2, 3
368
4661631
DA530258.1
-
2, 3
422
DA773983.1
-
3, 4
549
DB127917.1
-
2, 3
393
DB155885.1
-
2, 3
462
Back To Top
More Information
Panther Classification:
Molecular Function -
SNARE protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of insulin secretion involved in cellular response to glucose stimulus
protein binding
microtubule binding
syntaxin-1 binding
kinesin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0