Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RIMKLB SIRNA Assays ›
          Gene Symbol:
          RIMKLB
          Gene Name:
          ribosomal modification protein rimK like family member B
          Gene Aliases:
          FAM80B, NAAGS, NAAGS-I
          Chromosome Location:
          Chr.12: 8681606 - 8783098 on Build GRCh38
          UniGene ID:
          Hs.504670
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 258572

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RIMKLB
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001297776.1 NP_001284705.1 2 515
          NM_020734.3 NP_065785.2 2 947
          NR_123740.1 2 259
          XM_006719118.2 XP_006719181.1 2 636
          XM_006719127.2 XP_006719190.1 2 636
          XM_011520776.1 XP_011519078.1 2 636
          XM_011520777.1 XP_011519079.1 2 636
          XM_011520778.2 XP_011519080.1 2 636
          XM_011520779.2 XP_011519081.1 2 636
          XM_011520780.1 XP_011519082.1 2 636
          XM_011520783.1 XP_011519085.1 2 636
          XM_011520790.1 XP_011519092.1 3 1323
          XM_017019677.1 XP_016875166.1 2 636
          XM_017019678.1 XP_016875167.1 2 636
          XM_017019679.1 XP_016875168.1 2 636
          XM_017019680.1 XP_016875169.1 2 636
          XM_017019681.1 XP_016875170.1 2 636
          XM_017019682.1 XP_016875171.1 2 515
          XM_017019683.1 XP_016875172.1 3 1323
          XM_017019684.1 XP_016875173.1 2 259
          XM_017019685.1 XP_016875174.1 3 1038
          XM_017019686.1 XP_016875175.1 2 273
          XM_017019687.1 XP_016875176.1 3 637
          XM_017019688.1 XP_016875177.1 2 777
          XM_017019689.1 XP_016875178.1 2 326
          XM_017019690.1 XP_016875179.1 2 137
          XM_017019691.1 XP_016875180.1 2 515
          XM_017019692.1 XP_016875181.1 2 493
          XM_017019693.1 XP_016875182.1 2 515
          XM_017019694.1 XP_016875183.1 2 520
          XM_017019695.1 XP_016875184.1 2 289
          XM_017019696.1 XP_016875185.1 3 1323
          XM_017019697.1 XP_016875186.1 2 210
          GenBank mRNA AB033064.1 - 2 886
          AK302847.1 - 2 515
          BC006368.2 - 3 1208
          BC033793.1 - 2 256 5139256
          DB265022.1 - 2 259

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular protein modification process
          ATP binding
          metal ion binding
          N-acetyl-L-aspartate-L-glutamate ligase activity
          citrate-L-glutamate ligase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0