Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MACF1 SIRNA Assays ›
          Gene Symbol:
          MACF1
          Gene Name:
          microtubule-actin crosslinking factor 1
          Gene Aliases:
          ABP620, ACF7, MACF, OFC4
          Chromosome Location:
          Chr.1: 39084167 - 39487138 on Build GRCh38
          UniGene ID:
          Hs.472475
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 258846

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MACF1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012090.5 NP_036222.3 36, 37 4824
          XM_005270694.1 XP_005270751.1 37, 38 11686
          XM_005270695.1 XP_005270752.1 37, 38 11686
          XM_005270696.1 XP_005270753.1 37, 38 11686
          XM_005270697.1 XP_005270754.1 37, 38 11686
          XM_005270698.1 XP_005270755.1 37, 38 11686
          XM_006710519.1 XP_006710582.1 37, 38 11686
          XM_006710520.2 XP_006710583.1 37, 38 11686
          XM_006710521.1 XP_006710584.1 37, 38 11686
          XM_006710522.1 XP_006710585.1 37, 38 11686
          XM_006710524.2 XP_006710587.1 36, 37 11452
          XM_006710526.3 XP_006710589.1 42, 43 11118
          XM_006710528.3 XP_006710591.1 42, 43 11679
          XM_006710529.3 XP_006710592.1 42, 43 11036
          XM_006710530.3 XP_006710593.1 42, 43 11924
          XM_006710531.3 XP_006710594.1 42, 43 11655
          XM_006710532.3 XP_006710595.1 41, 42 11583
          XM_006710533.2 XP_006710596.1 41, 42 11583
          XM_006710535.3 XP_006710598.1 42, 43 11188
          XM_006710536.2 XP_006710599.1 32, 33 5485
          XM_006710538.2 XP_006710601.1 37, 38 11686
          XM_011541141.2 XP_011539443.1 37, 38 11632
          XM_011541142.2 XP_011539444.1 36, 37 6235
          XM_017000873.1 XP_016856362.1 37, 38 11686
          XM_017000874.1 XP_016856363.1 37, 38 11686
          XM_017000875.1 XP_016856364.1 41, 42 11008
          XM_017000876.1 XP_016856365.1 41, 42 11092
          XM_017000877.1 XP_016856366.1 42, 43 11014
          XM_017000878.1 XP_016856367.1 42, 43 10877
          XM_017000879.1 XP_016856368.1 7, 8 6448
          XM_017000880.1 XP_016856369.1 32, 33 5485
          XM_017000881.1 XP_016856370.1 36, 37 4799
          XM_017000882.1 XP_016856371.1 32, 33 5485
          GenBank mRNA AB007934.3 - 16, 17 2172
          AB029290.1 - 36, 37 4823
          AB033077.1 - 16, 17 2172
          AF317696.1 - 6, 7 6398

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001135758, XM_008764041, XM_008764038, XM_008764034, XM_008764044, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001199136, NM_001199137, AK220378.

          Panther Classification:

          Molecular Function -

          intermediate filament binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of epithelial cell migration
          Wnt signaling pathway
          positive regulation of Wnt signaling pathway
          regulation of microtubule-based process
          wound healing
          Golgi to plasma membrane protein transport
          positive regulation of axon extension
          regulation of focal adhesion assembly
          cell-cell adhesion
          actin binding
          calcium ion binding
          protein binding
          microtubule binding
          ATPase activity
          poly(A) RNA binding
          actin filament binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0