Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PIK3R6 SIRNA Assays ›
          Gene Symbol:
          PIK3R6
          Gene Name:
          phosphoinositide-3-kinase regulatory subunit 6
          Gene Aliases:
          C17orf38, HsT41028, p84 PIKAP, p87(PIKAP), p87PIKAP
          Chromosome Location:
          Chr.17: 8802723 - 8870003 on Build GRCh38
          UniGene ID:
          Hs.255809
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 259211

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PIK3R6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001010855.3 NP_001010855.1 19 2324
          NM_001290211.1 NP_001277140.1 19 2692
          NR_110865.1 18 2419
          XM_011523677.2 XP_011521979.1 19 2218
          XM_011523678.2 XP_011521980.1 19 2205
          XM_011523680.2 XP_011521982.1 18 2098
          XM_011523681.2 XP_011521983.1 18 2469
          XM_011523683.2 XP_011521985.1 16 1906
          XM_011523686.2 XP_011521988.1 10 1355
          XM_017024228.1 XP_016879717.1 19 2220
          XM_017024229.1 XP_016879718.1 20 2336
          XM_017024230.1 XP_016879719.1 19 2213
          XM_017024231.1 XP_016879720.1 15 2397
          GenBank mRNA AK091819.1 - 18 2421
          AY753192.1 - 17 2084
          BC110301.1 - 19 2675
          BC125169.1 - 14 1728

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          kinase modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          phosphatidylinositol biosynthetic process
          G-protein coupled receptor signaling pathway
          platelet activation
          regulation of natural killer cell mediated cytotoxicity
          positive regulation of MAP kinase activity
          regulation of phosphatidylinositol 3-kinase activity
          positive regulation of T cell differentiation
          positive regulation of angiogenesis
          phosphatidylinositol phosphorylation
          protein binding
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity
          1-phosphatidylinositol-3-kinase regulator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0