Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FGFR2 SIRNA Assays ›
          Gene Symbol:
          Fgfr2
          Gene Name:
          fibroblast growth factor receptor 2
          Gene Aliases:
          AU043015, AW556123, Bek, Fgfr-2, Fgfr-7, Fgfr7, KGFR, KGFRTr, svs
          Chromosome Location:
          Chr.7: 130162451 - 130266944 on Build GRCm38
          UniGene ID:
          Mm.16340
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 261055

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Fgfr2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_010207.2 NP_034337.2 11 2730
          NM_201601.2 NP_963895.2 9 2388
          GenBank mRNA BC065069.1 - 9 1810
          BC072640.1 - 10 2022
          BC151201.1 - 9 1590 9088089
          BC172174.1 - 10 1671 9054844
          EF143322.1 - 9 1242
          EF143323.1 - 10 1509
          EF143324.1 - 8 1164
          EF143325.1 - 9 1236
          EF143326.1 - 9 1242
          EF143327.1 - 8 1158
          EF143328.1 - 10 1503
          EF143329.1 - 10 1509
          EF143330.1 - 8 1053
          EF143331.1 - 8 1158
          EF143332.1 - 10 1387
          EF143333.1 - 6 867
          EF143334.1 - 8 1155
          EF143335.1 - 10 1500
          EF143336.1 - 8 1164
          EF143337.1 - 6 870
          EF143338.1 - 10 1506
          EF143339.1 - 8 1155
          EF143340.1 - 8 1161
          M23362.1 - 1 78
          M63503.1 - 9 1839
          M86441.1 - 11 2112
          X55441.1 - 11 2160

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          angiogenesis
          ureteric bud development
          in utero embryonic development
          epithelial to mesenchymal transition
          positive regulation of mesenchymal cell proliferation
          outflow tract septum morphogenesis
          membranous septum morphogenesis
          protein phosphorylation
          apoptotic process
          cell-cell signaling
          axonogenesis
          neuromuscular junction development
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          fibroblast growth factor receptor signaling pathway
          regulation of smoothened signaling pathway
          post-embryonic development
          embryonic pattern specification
          organ morphogenesis
          regulation of cell fate commitment
          positive regulation of phospholipase activity
          phosphorylation
          morphogenesis of embryonic epithelium
          peptidyl-tyrosine phosphorylation
          orbitofrontal cortex development
          ventricular zone neuroblast division
          pyramidal neuron development
          gland morphogenesis
          positive regulation of Wnt signaling pathway
          bone mineralization
          lung development
          epithelial cell differentiation
          midbrain development
          otic vesicle formation
          hair follicle morphogenesis
          lacrimal gland development
          multicellular organism growth
          organ growth
          fibroblast growth factor receptor signaling pathway involved in negative regulation of apoptotic process in bone marrow
          fibroblast growth factor receptor signaling pathway involved in hemopoiesis
          fibroblast growth factor receptor signaling pathway involved in positive regulation of cell proliferation in bone marrow
          fibroblast growth factor receptor signaling pathway involved in orbitofrontal cortex development
          regulation of multicellular organism growth
          regulation of fibroblast growth factor receptor signaling pathway
          regulation of cell proliferation
          inner ear morphogenesis
          odontogenesis
          positive regulation of MAPK cascade
          cell fate commitment
          positive regulation of cell cycle
          negative regulation of mitotic nuclear division
          positive regulation of transcription from RNA polymerase II promoter
          protein autophosphorylation
          lung alveolus development
          synaptic vesicle transport
          embryonic digestive tract morphogenesis
          embryonic organ morphogenesis
          digestive tract development
          embryonic organ development
          reproductive structure development
          positive regulation of smooth muscle cell proliferation
          embryonic cranial skeleton morphogenesis
          epidermis morphogenesis
          branching morphogenesis of a nerve
          mesenchymal cell differentiation
          epithelial cell proliferation
          regulation of epithelial cell proliferation
          positive regulation of epithelial cell proliferation
          negative regulation of epithelial cell proliferation
          regulation of smooth muscle cell differentiation
          positive regulation of cell division
          ventricular cardiac muscle tissue morphogenesis
          positive regulation of cardiac muscle cell proliferation
          limb bud formation
          bone development
          bone morphogenesis
          coronal suture morphogenesis
          epithelial tube branching involved in lung morphogenesis
          branching involved in prostate gland morphogenesis
          branching involved in salivary gland morphogenesis
          bud elongation involved in lung branching
          lung lobe morphogenesis
          lung-associated mesenchyme development
          positive regulation of epithelial cell proliferation involved in lung morphogenesis
          prostate gland morphogenesis
          prostate epithelial cord elongation
          prostate epithelial cord arborization involved in prostate glandular acinus morphogenesis
          squamous basal epithelial stem cell differentiation involved in prostate gland acinus development
          fibroblast growth factor receptor signaling pathway involved in mammary gland specification
          lateral sprouting from an epithelium
          mammary gland bud formation
          epithelial cell proliferation involved in salivary gland morphogenesis
          branch elongation involved in salivary gland morphogenesis
          branching involved in labyrinthine layer morphogenesis
          regulation of branching involved in prostate gland morphogenesis
          regulation of morphogenesis of a branching structure
          mesenchymal cell differentiation involved in lung development
          mesenchymal cell proliferation involved in lung development
          vasculogenesis involved in coronary vascular morphogenesis
          endodermal digestive tract morphogenesis
          lens fiber cell development
          regulation of ERK1 and ERK2 cascade
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of canonical Wnt signaling pathway
          nucleotide binding
          protein kinase activity
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          fibroblast growth factor-activated receptor activity
          protein binding
          ATP binding
          heparin binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          fibroblast growth factor binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0