Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other OXT SIRNA Assays ›
Gene Symbol:
Oxt
Gene Name:
oxytocin/neurophysin 1 prepropeptide
Gene Aliases:
-
Chromosome Location:
Chr.3: 123106694 - 123107534 on Build Rnor_6.0
UniGene ID:
Rn.48915
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
261105
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Oxt
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012996.3
NP_037128.1
1, 2
157
GenBank mRNA
M25649.1
-
1, 2
157
Back To Top
More Information
Panther Classification:
Molecular Function -
neuropeptide
Gene Ontology Categories:
Function(s)
Process(es)
response to amphetamine
regulation of heart rate
maternal aggressive behavior
positive regulation of cytosolic calcium ion concentration
heart development
female pregnancy
parturition
lactation
memory
grooming behavior
response to mechanical stimulus
response to sucrose
positive regulation of norepinephrine secretion
response to organic cyclic compound
response to activity
sleep
response to food
positive regulation of prostaglandin secretion
response to estradiol
response to retinoic acid
response to progesterone
response to prostaglandin E
social behavior
negative regulation of urine volume
positive regulation of renal sodium excretion
response to cocaine
hyperosmotic salinity response
maternal behavior
sperm ejaculation
eating behavior
drinking behavior
response to external biotic stimulus
response to peptide hormone
regulation of digestive system process
response to ether
negative regulation of blood pressure
positive regulation of blood pressure
positive regulation of ossification
positive regulation of female receptivity
response to steroid hormone
positive regulation of synaptic transmission
response to glucocorticoid
response to cAMP
response to electrical stimulus
regulation of sensory perception of pain
positive regulation of synapse assembly
male mating behavior
positive regulation of penile erection
positive regulation of hindgut contraction
negative regulation of gastric acid secretion
positive regulation of uterine smooth muscle contraction
neuropeptide hormone activity
neurohypophyseal hormone activity
protein binding
oxytocin receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-wl2rl:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline