Search
Search

Epsti1| Gene Symbol: Epsti1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029495.2 | NP_083771.1 | 5 | 666 | ||
| NM_178825.4 | NP_849147.1 | 5 | 666 | |||
| GenBank mRNA | AK017174.1 | - | 5 | 499 | ||
| AK077223.1 | - | 5 | 472 | |||
| AK077376.1 | - | 5 | 501 | |||
| AK135710.1 | - | 5 | 660 | |||
| AK136055.1 | - | 5 | 667 | |||
| AY239223.1 | - | 5 | 499 | |||
| AY239224.1 | - | 5 | 572 | |||
| BC020120.1 | - | 5 | 499 | 3988503 | ||
| BC021821.1 | - | 5 | 526 | 3581940 | ||