Search
Search

Anks6| Gene Symbol: Anks6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024136.1 | NP_001019307.1 | 15 | 3363 | ||
| GenBank mRNA | AK008967.1 | - | 1 | 762 | ||
| AK085318.1 | - | 1 | 544 | |||
| BC060663.1 | - | 17 | 3771 | |||
| BC072562.1 | - | 9 | 1954 | |||