Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NPIPA3 SIRNA Assays ›
          Gene Symbol:
          NPIPA3 NPIPA2 NPIPA5 NPIPB9 PDXDC2P
          Gene Name:
          nuclear pore complex interacting protein family member A3
          nuclear pore complex interacting protein family member A2
          nuclear pore complex interacting protein family member A5
          nuclear pore complex interacting protein family member B9
          pyridoxal dependent decarboxylase domain containing 2, pseudogene
          Gene Aliases:
          -
          NPIP
          NPIP
          -
          NPIP, PDXDC2
          Chromosome Location:
          Chr.16: 14706473 - 14726338 on Build GRCh38
          Chromosome Location:
          Chr.16: 14745595 - 14765448 on Build GRCh38
          Chromosome Location:
          Chr.16: 15363624 - 15381478 on Build GRCh38
          Chromosome Location:
          Chr.16: 28735500 - 28772911 on Build GRCh38
          Chromosome Location:
          Chr.16: 69976297 - 70065948 on Build GRCh38
          UniGene ID:
          -
          -
          Hs.676266
          Hs.710214
          Hs.513695
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          siRNA ID 263661

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NPIPA3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_011522576.2 XP_011520878.2 2, 3 2433
          XM_011522577.2 XP_011520879.2 2, 3 2354
          XM_011522581.1 XP_011520883.1 2, 3 2443
          XM_011522583.2 XP_011520885.1 2, 3 2443
          XM_011522584.1 XP_011520886.1 2, 3 2443
          XM_011522586.1 XP_011520888.1 2, 3 2443
          XM_017023553.1 XP_016879042.1 2, 3 2443
          Gene Symbol: NPIPA2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006720917.3 XP_006720980.2 2, 3 2443
          XM_011522587.1 XP_011520889.1 2, 3 2443
          XM_011522593.1 XP_011520895.1 2, 3 2443
          XM_011522594.2 XP_011520896.1 2, 3 2443
          XM_011522595.1 XP_011520897.1 2, 3 2443
          XM_011522597.1 XP_011520899.1 2, 3 2443
          Gene Symbol: NPIPA5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_011522329.2 XP_011520631.1 2, 3 391
          XM_011522336.2 XP_011520638.1 2, 3 384
          Gene Symbol: NPIPB9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_011545690.2 XP_011543992.1 2, 3 725
          XM_011545693.2 XP_011543995.1 2, 3 724
          XM_017022814.1 XP_016878303.1 2, 3 725
          XM_017022815.1 XP_016878304.1 2, 3 245
          XM_017022816.1 XP_016878305.1 2, 3 725
          XM_017022828.1 XP_016878317.1 2, 3 724
          Gene Symbol: PDXDC2P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003610.1 18, 19 1809
          GenBank mRNA AL833251.1 - 18, 19 1809
          BC063633.1 - 17, 18 1681

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          carboxylic acid metabolic process
          carboxy-lyase activity
          pyridoxal phosphate binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x9n5h:80/100.66.76.163:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0