Search
Search

TTC36| Gene Symbol: TTC36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080441.1 | NP_001073910.1 | 2 | 259 | ||
| GenBank mRNA | BC146916.1 | - | 2 | 282 | 9056844 | |
| BC146921.1 | - | 2 | 282 | 9056849 | ||
| BC171906.1 | - | 3 | 394 | 9054560 | ||
| BC171913.1 | - | 3 | 417 | 9054567 | ||
| EU489483.1 | - | 2 | 321 | |||