Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FGGY SIRNA Assays ›
          Gene Symbol:
          FGGY
          Gene Name:
          FGGY carbohydrate kinase domain containing
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 59296405 - 59762730 on Build GRCh38
          UniGene ID:
          Hs.444301
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 26600

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FGGY
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001113411.1 NP_001106882.1 5 712
          NM_001244714.1 NP_001231643.1 3 349
          NM_018291.3 NP_060761.3 5 712
          NR_103473.1 6 670
          XM_011541730.1 XP_011540032.1 5 636
          XM_011541731.1 XP_011540033.1 5 570
          XM_011541732.1 XP_011540034.1 5 613
          XM_011541733.1 XP_011540035.1 5 531
          XM_011541735.1 XP_011540037.1 5 531
          XM_011541736.2 XP_011540038.1 3 477
          XM_017001643.1 XP_016857132.1 5 742
          XM_017001644.1 XP_016857133.1 5 535
          XM_017001645.1 XP_016857134.1 5 636
          XM_017001646.1 XP_016857135.1 5 613
          XM_017001647.1 XP_016857136.1 5 741
          XM_017001648.1 XP_016857137.1 6 555
          XM_017001649.1 XP_016857138.1 5 681
          XM_017001650.1 XP_016857139.1 7 904
          XM_017001651.1 XP_016857140.1 7 760
          XM_017001652.1 XP_016857141.1 5 681
          XM_017001653.1 XP_016857142.1 5 613
          XM_017001654.1 XP_016857143.1 3 432
          XM_017001655.1 XP_016857144.1 3 341
          XM_017001656.1 XP_016857145.1 6 748
          XM_017001657.1 XP_016857146.1 5 536
          XM_017001658.1 XP_016857147.1 6 679
          XM_017001659.1 XP_016857148.1 4 569
          XM_017001660.1 XP_016857149.1 4 472
          XM_017001661.1 XP_016857150.1 4 629
          XM_017001662.1 XP_016857151.1 4 314
          XM_017001663.1 XP_016857152.1 4 489
          XM_017001664.1 XP_016857153.1 3 366
          XM_017001665.1 XP_016857154.1 3 273
          XM_017001666.1 XP_016857155.1 3 427
          XM_017001667.1 XP_016857156.1 3 481
          XM_017001668.1 XP_016857157.1 3 237
          XM_017001669.1 XP_016857158.1 3 332
          XM_017001670.1 XP_016857159.1 3 1105
          XM_017001671.1 XP_016857160.1 3 233
          XM_017001672.1 XP_016857161.1 4 569
          XM_017001673.1 XP_016857162.1 4 626
          XM_017001674.1 XP_016857163.1 4 485
          XM_017001675.1 XP_016857164.1 4 569
          XM_017001676.1 XP_016857165.1 4 629
          XM_017001679.1 XP_016857168.1 5 681
          GenBank mRNA AK022237.1 - 4 569
          AK054842.1 - 3 340
          AK090568.1 - 6 670
          AK315649.1 - 5 597
          BC014947.1 - 5 597 4871664
          BX647626.1 - 5 684
          DA969344.1 - 4 497

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001013932, XM_008763856, XM_006238413, XM_006238410, BC090077, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001113412, NM_029347, CB953569, BC031708, AK138439, and others.

          Panther Classification:

          Molecular Function -

          carbohydrate kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          phosphorylation
          neuron cellular homeostasis
          molecular_function
          kinase activity
          phosphotransferase activity, alcohol group as acceptor

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-lgjf9:80/100.66.79.224:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline