Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPP6R3 SIRNA Assays ›
Gene Symbol:
PPP6R3
Gene Name:
protein phosphatase 6 regulatory subunit 3
Gene Aliases:
C11orf23, PP6R3, SAP190, SAPL, SAPLa, SAPS3
Chromosome Location:
Chr.11: 68460712 - 68615334 on Build GRCh38
UniGene ID:
Hs.503022
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
26619
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPP6R3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164160.1
NP_001157632.1
3
350
NM_001164161.1
NP_001157633.1
3
350
NM_001164162.1
NP_001157634.1
3
350
NM_001164163.1
NP_001157635.1
3
350
NM_001164164.1
NP_001157636.1
4
455
NM_018312.4
NP_060782.2
4
455
XM_006718608.3
XP_006718671.1
4
466
XM_006718612.3
XP_006718675.1
4
466
XM_006718613.2
XP_006718676.1
4
466
XM_006718614.2
XP_006718677.1
4
466
XM_006718618.3
XP_006718681.1
4
466
XM_006718621.2
XP_006718684.1
4
466
XM_006718622.3
XP_006718685.1
4
466
XM_006718623.3
XP_006718686.1
4
449
XM_006718624.2
XP_006718687.1
4
466
XM_006718627.3
XP_006718690.1
4
466
XM_011545135.1
XP_011543437.1
4
466
XM_011545136.2
XP_011543438.1
3
349
XM_011545137.2
XP_011543439.1
4
449
XM_011545138.2
XP_011543440.1
5
667
XM_011545139.1
XP_011543441.1
4
466
XM_011545140.2
XP_011543442.1
4
466
XM_011545141.1
XP_011543443.1
4
466
XM_011545142.2
XP_011543444.1
4
466
XM_011545143.1
XP_011543445.1
4
466
XM_011545145.2
XP_011543447.1
4
466
XM_017017962.1
XP_016873451.1
3
304
XM_017017963.1
XP_016873452.1
4
390
XM_017017964.1
XP_016873453.1
4
466
XM_017017965.1
XP_016873454.1
3
349
XM_017017966.1
XP_016873455.1
4
552
XM_017017967.1
XP_016873456.1
4
466
XM_017017968.1
XP_016873457.1
3
349
XM_017017969.1
XP_016873458.1
4
466
XM_017017970.1
XP_016873459.1
3
349
XM_017017971.1
XP_016873460.1
4
466
XM_017017972.1
XP_016873461.1
3
322
XM_017017973.1
XP_016873462.1
4
449
XM_017017974.1
XP_016873463.1
4
466
XM_017017975.1
XP_016873464.1
4
466
XM_017017976.1
XP_016873465.1
3
349
XM_017017983.1
XP_016873472.1
4
469
XM_017017984.1
XP_016873473.1
3
286
GenBank mRNA
AB046778.2
-
3
298
AF264779.1
-
3
360
AF264780.1
-
3
360
AK315136.1
-
3
319
BC105933.1
-
2
218
40029064
BC105934.1
-
2
218
40029058
BC107599.1
-
2
218
40029065
BP314824.1
-
3
350
CN351527.1
-
4
424
CR749815.1
-
3
313
CR933658.1
-
3
313
CR936718.1
-
3
318
DB207620.1
-
4
466
DQ111954.2
-
1
83
Back To Top
More Information
Panther Classification:
Molecular Function -
phosphatase modulator
Gene Ontology Categories:
Function(s)
Process(es)
regulation of phosphoprotein phosphatase activity
COPII vesicle coating
protein binding
protein phosphatase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0