Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PPP6R3 SIRNA Assays ›
          Gene Symbol:
          PPP6R3
          Gene Name:
          protein phosphatase 6 regulatory subunit 3
          Gene Aliases:
          C11orf23, PP6R3, SAP190, SAPL, SAPLa, SAPS3
          Chromosome Location:
          Chr.11: 68460712 - 68615334 on Build GRCh38
          UniGene ID:
          Hs.503022
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 26807

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PPP6R3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001164160.1 NP_001157632.1 4 653
          NM_001164161.1 NP_001157633.1 4 653
          NM_001164162.1 NP_001157634.1 4 653
          NM_001164163.1 NP_001157635.1 4 653
          NM_001164164.1 NP_001157636.1 5 758
          NM_018312.4 NP_060782.2 5 758
          XM_006718608.3 XP_006718671.1 5 769
          XM_006718612.3 XP_006718675.1 5 769
          XM_006718613.2 XP_006718676.1 5 769
          XM_006718614.2 XP_006718677.1 5 769
          XM_006718618.3 XP_006718681.1 5 769
          XM_006718621.2 XP_006718684.1 5 769
          XM_006718622.3 XP_006718685.1 5 769
          XM_006718623.3 XP_006718686.1 5 752
          XM_006718624.2 XP_006718687.1 5 769
          XM_006718627.3 XP_006718690.1 5 769
          XM_011545135.1 XP_011543437.1 5 769
          XM_011545136.2 XP_011543438.1 4 652
          XM_011545137.2 XP_011543439.1 5 752
          XM_011545138.2 XP_011543440.1 6 970
          XM_011545139.1 XP_011543441.1 5 769
          XM_011545140.2 XP_011543442.1 5 769
          XM_011545141.1 XP_011543443.1 5 769
          XM_011545142.2 XP_011543444.1 5 769
          XM_011545143.1 XP_011543445.1 5 769
          XM_011545145.2 XP_011543447.1 5 769
          XM_017017962.1 XP_016873451.1 4 607
          XM_017017963.1 XP_016873452.1 5 693
          XM_017017964.1 XP_016873453.1 5 769
          XM_017017965.1 XP_016873454.1 4 652
          XM_017017966.1 XP_016873455.1 5 855
          XM_017017967.1 XP_016873456.1 5 769
          XM_017017968.1 XP_016873457.1 4 652
          XM_017017969.1 XP_016873458.1 5 769
          XM_017017970.1 XP_016873459.1 4 652
          XM_017017971.1 XP_016873460.1 5 769
          XM_017017972.1 XP_016873461.1 4 625
          XM_017017973.1 XP_016873462.1 5 752
          XM_017017974.1 XP_016873463.1 5 769
          XM_017017975.1 XP_016873464.1 5 769
          XM_017017976.1 XP_016873465.1 4 652
          XM_017017977.1 XP_016873466.1 4 540
          XM_017017978.1 XP_016873467.1 3 391
          XM_017017979.1 XP_016873468.1 3 391
          XM_017017980.1 XP_016873469.1 3 391
          XM_017017981.1 XP_016873470.1 3 391
          XM_017017982.1 XP_016873471.1 3 391
          XM_017017983.1 XP_016873472.1 5 772
          XM_017017984.1 XP_016873473.1 4 589
          GenBank mRNA AB046778.2 - 4 601
          AF264779.1 - 4 663
          AF264780.1 - 4 663
          AK298180.1 - 4 518
          AK303315.1 - 3 392
          AK315136.1 - 4 622
          BC105933.1 - 3 521 40029064
          BC105934.1 - 3 521 40029058
          BC107599.1 - 3 521 40029065
          CR749815.1 - 4 616
          CR933658.1 - 4 616
          CR936718.1 - 4 621
          DQ111954.2 - 2 386

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001287134, XM_008760130, XM_008760124, XM_008760122, XM_008760125, and others.
          This siRNA also targets the following transcripts in Mouse: NM_029456, NM_028999, NM_001164159, AK088526, BC008529, and others.

          Panther Classification:

          Molecular Function -

          phosphatase modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of phosphoprotein phosphatase activity
          COPII vesicle coating
          protein binding
          protein phosphatase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0