Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TBC1D3L SIRNA Assays ›
          Gene Symbol:
          TBC1D3L LOC101060389 TBC1D3C TBC1D3H TBC1D3I TBC1D3K
          Gene Name:
          TBC1 domain family member 3L
          TBC1 domain family member-like
          TBC1 domain family member 3C
          TBC1 domain family member 3H
          TBC1 domain family member 3I
          TBC1 domain family member 3K
          Gene Aliases:
          -
          TBC1D3D
          TBC1D3D
          -
          -
          -
          Chromosome Location:
          Chr.17: 37978155 - 37990691 on Build GRCh38
          Chromosome Location:
          Chr.17: 38004033 - 38014908 on Build GRCh38
          Chromosome Location:
          Chr.17: 38057693 - 38068634 on Build GRCh38
          Chromosome Location:
          Chr.17: 36377475 - 36388448 on Build GRCh38
          Chromosome Location:
          Chr.17: 36253639 - 36264601 on Build GRCh38
          Chromosome Location:
          Chr.17: 37923805 - 37935371 on Build GRCh38
          UniGene ID:
          Hs.454716
          Hs.454716
          Hs.735975
          Hs.105891
          Hs.454716
          Hs.454716
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          siRNA ID 270007

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TBC1D3L
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006722242.3 XP_006722305.1 1 2936
          XM_006722243.2 XP_006722306.1 1 1188
          XM_006722244.2 XP_006722307.1 1 1188
          XM_006722246.3 XP_006722309.1 1 519
          XM_006722247.2 XP_006722310.1 1 1188
          GenBank mRNA AK095385.1 - 1 519
          AX775949.1 - 1 519
          AX775951.1 - 1 519
          Gene Symbol: LOC101060389
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006722248.2 XP_006722311.1 1 1188
          XM_006722249.2 XP_006722312.1 1 1188
          XM_006722250.2 XP_006722313.1 1 1188
          XM_006722252.3 XP_006722315.1 1 1188
          XM_006722253.2 XP_006722316.1 1 1188
          XM_017024010.1 XP_016879499.1 1 1188
          Gene Symbol: TBC1D3C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006721904.2 XP_006721967.1 1 1188
          XM_006721905.2 XP_006721968.1 1 1188
          XM_006721907.3 XP_006721970.1 1 1188
          XM_006721908.2 XP_006721971.1 1 1188
          XM_017024646.1 XP_016880135.1 1 1188
          Gene Symbol: TBC1D3H
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006722055.1 XP_006722118.1 1 519
          XM_006722056.1 XP_006722119.1 1 519
          XM_011525172.1 XP_011523474.1 1 519
          XM_011525174.1 XP_011523476.1 1 519
          XM_011525175.1 XP_011523477.1 1 519
          XM_011525176.1 XP_011523478.1 1 519
          Gene Symbol: TBC1D3I
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006722224.3 XP_006722287.1 1 1381
          XM_006722226.3 XP_006722289.1 1 1381
          XM_011524183.2 XP_011522485.1 1 1381
          XM_011524184.2 XP_011522486.1 1 1381
          XM_011524185.2 XP_011522487.1 1 1381
          Gene Symbol: TBC1D3K
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006722235.2 XP_006722298.1 1 1960
          XM_006722237.2 XP_006722300.1 1 1960
          XM_006722239.2 XP_006722302.1 1 1960
          XM_011524172.1 XP_011522474.1 1 1960
          XM_011524173.1 XP_011522475.1 1 1960
          XM_011524174.1 XP_011522476.1 1 1960

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of GTPase activity
          intracellular protein transport
          regulation of vesicle fusion
          activation of GTPase activity
          GTPase activator activity
          Rab GTPase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0