Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PCID2 SIRNA Assays ›
Gene Symbol:
PCID2
Gene Name:
PCI domain containing 2
Gene Aliases:
F10
Chromosome Location:
Chr.13: 113165002 - 113209466 on Build GRCh38
UniGene ID:
Hs.508769
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
27059
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PCID2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001127202.3
NP_001120674.1
14
1809
NM_001127203.3
NP_001120675.1
15
1644
NM_001258213.2
NP_001245142.1
14
2112
NM_001320655.1
NP_001307584.1
15
1947
NM_001320656.1
NP_001307585.1
15
1774
NM_001320657.1
NP_001307586.1
14
1813
NM_001320659.1
NP_001307588.1
15
1648
NM_001320660.1
NP_001307589.1
14
2116
NM_018386.4
NP_060856.2
15
1612
XM_017020664.1
XP_016876153.1
15
1363
XM_017020665.1
XP_016876154.1
14
2118
XM_017020666.1
XP_016876155.1
14
1613
XM_017020667.1
XP_016876156.1
13
2251
XM_017020668.1
XP_016876157.1
13
1456
XM_017020669.1
XP_016876158.1
13
2297
XM_017020670.1
XP_016876159.1
8
2021
XM_017020671.1
XP_016876160.1
8
1944
XM_017020672.1
XP_016876161.1
9
1779
GenBank mRNA
AF183426.1
-
15
1387
AK000888.1
-
12
1507
AK001185.1
-
14
1567
AK001302.1
-
15
1339
AK002167.1
-
15
1523
AK023313.1
-
15
1359
AK024478.1
-
8
3931
AK098706.1
-
15
1405
AK289462.1
-
15
1366
BC016614.1
-
15
1390
3869275
BC031246.1
-
14
2114
5266649
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of apoptotic process
regulation of mRNA stability
positive regulation of B cell differentiation
positive regulation of transcription, DNA-templated
spleen development
positive regulation of mitotic cell cycle spindle assembly checkpoint
negative regulation of cysteine-type endopeptidase activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0