Search
Search

CARNS1| Gene Symbol: CARNS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166222.1 | NP_001159694.1 | 10 | 1797 | ||
| NM_020811.1 | NP_065862.1 | 9 | 1770 | |||
| XM_011545189.2 | XP_011543491.1 | 9 | 2087 | |||
| XM_011545191.2 | XP_011543493.1 | 8 | 4445 | |||
| XM_017018054.1 | XP_016873543.1 | 9 | 3001 | |||
| XM_017018055.1 | XP_016873544.1 | 7 | 2281 | |||
| GenBank mRNA | AB037815.1 | - | 8 | 2895 | ||
| AK289938.1 | - | 1 | 133 | |||
| AK294035.1 | - | 11 | 1719 | |||
| BC036557.1 | - | 1 | 168 | |||
| EF560746.1 | - | 9 | 1775 | |||