Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PLB1 SIRNA Assays ›
Gene Symbol:
PLB1
Gene Name:
phospholipase B1
Gene Aliases:
PLB, PLB/LIP, hPLB
Chromosome Location:
Chr.2: 28488579 - 28644135 on Build GRCh38
UniGene ID:
Hs.444933
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
272612
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PLB1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001170585.1
NP_001164056.1
31
2222
NM_153021.4
NP_694566.4
32
2255
XM_011532579.2
XP_011530881.1
34
2562
XM_011532581.2
XP_011530883.1
34
2525
XM_011532584.2
XP_011530886.1
34
2562
XM_011532588.2
XP_011530890.1
34
2564
XM_011532589.2
XP_011530891.1
34
2559
XM_011532590.2
XP_011530892.1
34
2564
XM_011532591.2
XP_011530893.1
34
2561
XM_011532593.2
XP_011530895.1
30
2137
XM_011532601.2
XP_011530903.1
20
1550
XM_011532602.2
XP_011530904.1
19
1522
XM_011532603.2
XP_011530905.1
19
1599
XM_011532606.2
XP_011530908.1
34
2584
XM_011532607.2
XP_011530909.1
34
2584
XM_011532608.2
XP_011530910.1
34
2582
XM_011532609.2
XP_011530911.1
17
1353
XM_011532610.2
XP_011530912.1
9
1539
XM_017003432.1
XP_016858921.1
33
2531
XM_017003433.1
XP_016858922.1
33
2497
XM_017003434.1
XP_016858923.1
34
2566
GenBank mRNA
AK055428.1
-
13
938
BC065041.1
-
19
1341
BC150633.1
-
32
2255
9057003
BC153864.1
-
31
2174
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_138898, D63648.
Panther Classification:
Molecular Function -
hydrolase
lipase
phospholipase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
retinoid metabolic process
lipid catabolic process
phosphatidylcholine acyl-chain remodeling
retinol metabolic process
positive regulation of acrosome reaction
lysophospholipase activity
phospholipase A2 activity
retinyl-palmitate esterase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-rhr4g:80/100.66.78.163:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0