Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMEM234 SIRNA Assays ›
Gene Symbol:
TMEM234
Gene Name:
transmembrane protein 234
Gene Aliases:
AASL548, C1orf91, PRO1105, RP4-622L5, dJ622L5.7
Chromosome Location:
Chr.1: 32204807 - 32222371 on Build GRCh38
UniGene ID:
Hs.272299
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
27757
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMEM234
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_019118.4
NP_061991.3
2
195
NR_133634.1
2
195
NR_133635.1
2
195
NR_133636.1
2
195
XM_011541795.2
XP_011540097.1
2
177
XM_017001812.1
XP_016857301.1
2
177
XM_017001813.1
XP_016857302.1
2
177
XM_017001814.1
XP_016857303.1
2
177
XM_017001815.1
XP_016857304.1
2
177
XM_017001816.1
XP_016857305.1
2
177
XM_017001817.1
XP_016857306.1
2
177
XM_017001818.1
XP_016857307.1
2
177
XM_017001819.1
XP_016857308.1
2
177
XM_017001820.1
XP_016857309.1
2
177
XM_017001821.1
XP_016857310.1
2
177
XM_017001822.1
XP_016857311.1
2
177
XM_017001823.1
XP_016857312.1
2
177
XM_017001824.1
XP_016857313.1
2
177
XM_017001825.1
XP_016857314.1
2
177
XM_017001826.1
XP_016857315.1
2
177
XM_017001827.1
XP_016857316.1
2
177
XM_017001828.1
XP_016857317.1
2
177
XM_017001829.1
XP_016857318.1
2
177
XM_017001830.1
XP_016857319.1
2
177
GenBank mRNA
AF258548.1
-
2
172
AK295232.1
-
2
195
AK312046.1
-
2
170
AY358586.1
-
2
173
BC001256.1
-
2
157
BC004885.2
-
2
149
3837394
BC038842.1
-
2
172
5742036
BP307734.1
-
2
165
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0