Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SHH SIRNA Assays ›
Gene Symbol:
SHH
Gene Name:
sonic hedgehog
Gene Aliases:
HHG1, HLP3, HPE3, MCOPCB5, SMMCI, TPT, TPTPS
Chromosome Location:
Chr.7: 155799984 - 155812273 on Build GRCh38
UniGene ID:
Hs.164537
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
2792
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SHH
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000193.3
NP_000184.1
1
341
GenBank mRNA
BX452210.1
-
1
246
L38518.1
-
1
341
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
patterning of blood vessels
vasculogenesis
metanephros development
branching involved in ureteric bud morphogenesis
cell fate specification
neural crest cell migration
heart looping
positive regulation of neuroblast proliferation
osteoblast development
lymphoid progenitor cell differentiation
determination of left/right asymmetry in lateral mesoderm
endocytosis
smoothened signaling pathway
positive regulation of hh target transcription factor activity
cell-cell signaling
pattern specification process
ectoderm development
neuroblast proliferation
axon guidance
central nervous system development
ventral midline development
hindgut morphogenesis
heart development
blood coagulation
androgen metabolic process
positive regulation of cell proliferation
embryo development
embryonic pattern specification
polarity specification of anterior/posterior axis
dorsal/ventral pattern formation
oligodendrocyte development
striated muscle tissue development
positive regulation of skeletal muscle cell proliferation
myotube differentiation
intein-mediated protein splicing
spinal cord dorsal/ventral patterning
spinal cord motor neuron differentiation
thalamus development
dorsal/ventral neural tube patterning
cerebellar granule cell precursor proliferation
smoothened signaling pathway involved in regulation of cerebellar granule cell precursor cell proliferation
telencephalon regionalization
establishment of cell polarity
regulation of proteolysis
positive regulation of Wnt signaling pathway
lung development
embryonic limb morphogenesis
negative regulation of cell migration
male genitalia development
prostate gland development
thyroid gland development
forebrain development
midbrain development
hindbrain development
pancreas development
hair follicle morphogenesis
negative regulation of proteasomal ubiquitin-dependent protein catabolic process
T cell differentiation in thymus
positive regulation of T cell differentiation in thymus
positive regulation of immature T cell proliferation in thymus
negative regulation of transcription elongation from RNA polymerase II promoter
protein localization to nucleus
embryonic forelimb morphogenesis
embryonic hindlimb morphogenesis
regulation of cell proliferation
negative regulation of T cell proliferation
positive regulation of protein import into nucleus
odontogenesis of dentin-containing tooth
regulation of odontogenesis
embryonic digit morphogenesis
camera-type eye development
negative regulation of apoptotic process
CD4-positive or CD8-positive, alpha-beta T cell lineage commitment
positive thymic T cell selection
negative thymic T cell selection
intermediate filament organization
myoblast differentiation
negative regulation of cell differentiation
positive regulation of smoothened signaling pathway
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of alpha-beta T cell differentiation
negative regulation of alpha-beta T cell differentiation
cell development
thymus development
embryonic digestive tract morphogenesis
embryonic foregut morphogenesis
positive regulation of skeletal muscle tissue development
organ formation
neuron fate commitment
embryonic skeletal system development
positive regulation of oligodendrocyte differentiation
branching morphogenesis of an epithelial tube
inner ear development
formation of anatomical boundary
stem cell development
positive regulation of striated muscle cell differentiation
positive regulation of cell division
Bergmann glial cell differentiation
palate development
canonical Wnt signaling pathway
limb bud formation
lung epithelium development
trachea morphogenesis
branching involved in salivary gland morphogenesis
bud outgrowth involved in lung branching
right lung development
left lung development
lung lobe morphogenesis
lung-associated mesenchyme development
primary prostatic bud elongation
prostate epithelial cord elongation
salivary gland cavitation
epithelial cell proliferation involved in salivary gland morphogenesis
regulation of prostatic bud formation
epithelial-mesenchymal signaling involved in prostate gland development
positive regulation of epithelial cell proliferation involved in prostate gland development
regulation of mesenchymal cell proliferation involved in prostate gland development
mesenchymal smoothened signaling pathway involved in prostate gland development
artery development
mesenchymal cell proliferation involved in lung development
somite development
positive regulation of sclerotome development
cellular response to lithium ion
dopaminergic neuron differentiation
renal system development
metanephric mesenchymal cell proliferation involved in metanephros development
multicellular structure septum development
negative regulation of canonical Wnt signaling pathway
negative regulation of cholesterol efflux
apoptotic signaling pathway
regulation of nodal signaling pathway involved in determination of lateral mesoderm left/right asymmetry
regulation of protein localization to nucleus
negative regulation of dopaminergic neuron differentiation
negative regulation of ureter smooth muscle cell differentiation
positive regulation of ureter smooth muscle cell differentiation
negative regulation of kidney smooth muscle cell differentiation
positive regulation of kidney smooth muscle cell differentiation
positive regulation of mesenchymal cell proliferation involved in ureter development
negative regulation of mesenchymal cell apoptotic process
glycoprotein binding
patched binding
calcium ion binding
protein binding
glycosaminoglycan binding
peptidase activity
zinc ion binding
morphogen activity
laminin-1 binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0