Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NPIPB9 SIRNA Assays ›
          Gene Symbol:
          NPIPB9 LOC105376752 NPIPA1 NPIPA2 NPIPA3 NPIPA5 NPIPA7 NPIPA8 NPIPB3 NPIPB8 PDXDC2P PKD1P1
          Gene Name:
          nuclear pore complex interacting protein family member B9
          nuclear pore complex-interacting protein family member A5
          nuclear pore complex interacting protein family member A1
          nuclear pore complex interacting protein family member A2
          nuclear pore complex interacting protein family member A3
          nuclear pore complex interacting protein family member A5
          nuclear pore complex interacting protein family member A7
          nuclear pore complex interacting protein family member A8
          nuclear pore complex interacting protein family member B3
          nuclear pore complex interacting protein family member B8
          pyridoxal dependent decarboxylase domain containing 2, pseudogene
          polycystin 1, transient receptor potential channel interacting pseudogene 1
          Gene Aliases:
          -
          -
          NPIP, NPIPA, morpheus
          NPIP
          -
          NPIP
          -
          LCR16a9
          KIAA0220L, NPIP, NPIPB, NPIPB5, NPIPL3
          -
          NPIP, PDXDC2
          HG1, NPIP
          Chromosome Location:
          Chr.16: 28735500 - 28772911 on Build GRCh38
          Chromosome Location:
          Chr.16: 18359533 - 18372055 on Build GRCh38
          Chromosome Location:
          Chr.16: 14935258 - 14952074 on Build GRCh38
          Chromosome Location:
          Chr.16: 14745595 - 14765448 on Build GRCh38
          Chromosome Location:
          Chr.16: 14706473 - 14726338 on Build GRCh38
          Chromosome Location:
          Chr.16: 15363624 - 15381478 on Build GRCh38
          Chromosome Location:
          Chr.16: 16376940 - 16393972 on Build GRCh38
          Chromosome Location:
          Chr.16: 18317919 - 18334967 on Build GRCh38
          Chromosome Location:
          Chr.16: 21402134 - 21425337 on Build GRCh38
          Chromosome Location:
          Chr.16: 28638065 - 28658693 on Build GRCh38
          Chromosome Location:
          Chr.16: 69976297 - 70065948 on Build GRCh38
          Chromosome Location:
          Chr.16: 16317609 - 16350608 on Build GRCh38
          UniGene ID:
          Hs.710214
          -
          Hs.626677
          -
          -
          Hs.676266
          Hs.634612
          Hs.636569
          Hs.611072
          -
          Hs.513695
          Hs.636569
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID 279732

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NPIPB9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_017022814.1 XP_016878303.1 8 1435
          XM_017022815.1 XP_016878304.1 8 955
          XM_017022816.1 XP_016878305.1 8 1435
          XM_017022817.1 XP_016878306.1 7 1347
          XM_017022818.1 XP_016878307.1 7 2672
          XM_017022819.1 XP_016878308.1 8 1362
          XM_017022820.1 XP_016878309.1 8 1079
          XM_017022821.1 XP_016878310.1 7 1945
          XM_017022822.1 XP_016878311.1 8 2948
          XM_017022823.1 XP_016878312.1 6 2584
          XM_017022826.1 XP_016878315.1 7 896
          XM_017022827.1 XP_016878316.1 3 643
          Gene Symbol: LOC105376752
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_011522797.2 XP_011521099.1 6 716
          Gene Symbol: NPIPA1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK294177.1 - 3 170
          Gene Symbol: NPIPA2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_011522587.1 XP_011520889.1 9 3156
          XM_011522596.1 XP_011520898.1 6 612
          Gene Symbol: NPIPA3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_011522585.1 XP_011520887.1 6 612
          Gene Symbol: NPIPA5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_011522329.2 XP_011520631.1 9 1104
          XM_011522337.1 XP_011520639.1 6 612
          Gene Symbol: NPIPA7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_017022840.1 XP_016878329.1 7 706
          Gene Symbol: NPIPA8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_017022847.1 XP_016878336.1 7 706
          GenBank mRNA BY996642.1 - 3 274
          Gene Symbol: NPIPB3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK308629.1 - 1 421
          Gene Symbol: NPIPB8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_017023622.1 XP_016879111.1 7 871
          XM_017023623.1 XP_016879112.1 7 871
          Gene Symbol: PDXDC2P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC144350.1 - 6 642 9052870
          BX647358.1 - 2 4017
          Gene Symbol: PKD1P1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK298381.1 - 5 451

          Back To Top

          More Information


          Gene Ontology Categories:

          Process(es)

          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0