Search
Search

Cox6a1| Gene Symbol: Cox6a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012814.1 | NP_036946.1 | 3 | 463 | ||
| GenBank mRNA | AA685532.1 | - | 1 | 170 | ||
| CB716915.1 | - | 3 | 463 | |||
| FQ211611.1 | - | 3 | 450 | |||
| FQ213920.1 | - | 3 | 450 | |||
| FQ221129.1 | - | 3 | 455 | |||
| FQ221354.1 | - | 3 | 450 | |||
| FQ221968.1 | - | 3 | 452 | |||
| FQ225833.1 | - | 3 | 450 | |||
| X12553.1 | - | 3 | 364 | |||