Search
Search

Rmdn2| Gene Symbol: Rmdn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037200.1 | NP_001032277.1 | 2 | 658 | ||
| XM_006239633.2 | XP_006239695.1 | 1 | 294 | |||
| XM_006239634.2 | XP_006239696.1 | 1 | 294 | |||
| XM_006239635.2 | XP_006239697.1 | 4 | 1122 | |||
| XM_008764422.1 | XP_008762644.1 | 1 | 294 | |||
| XM_008764423.1 | XP_008762645.1 | 4 | 1140 | |||
| GenBank mRNA | BC100261.1 | - | 2 | 658 | 7368645 | |