Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IGF1 SIRNA Assays ›
          Gene Symbol:
          Igf1
          Gene Name:
          insulin-like growth factor 1
          Gene Aliases:
          C730016P09Rik, Igf-1, Igf-I
          Chromosome Location:
          Chr.10: 87858270 - 87937047 on Build GRCm38
          UniGene ID:
          Mm.268521
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 282797

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Igf1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001111274.1 NP_001104744.1 4 651
          NM_010512.4 NP_034642.2 4 733
          NM_184052.3 NP_908941.1 4 651
          GenBank mRNA AK038119.1 - 4 1734
          AY878193.1 - 4 531
          BC012409.1 - 3 388 4194295
          CT010364.1 - 3 361
          X04482.1 - 4 433

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          cell activation
          regulation of protein phosphorylation
          blood vessel remodeling
          regulation of translation
          nervous system development
          memory
          cell proliferation
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          response to heat
          glial cell differentiation
          regulation of gene expression
          positive regulation of cardiac muscle hypertrophy
          phosphatidylinositol 3-kinase signaling
          positive regulation of phosphatidylinositol 3-kinase signaling
          skeletal muscle satellite cell maintenance involved in skeletal muscle regeneration
          muscle hypertrophy
          myotube cell development
          positive regulation of smooth muscle cell migration
          positive regulation of cerebellar granule cell precursor proliferation
          water homeostasis
          proteoglycan biosynthetic process
          positive regulation of cell growth
          lung development
          positive regulation of cell migration
          mammary gland development
          exocrine pancreas development
          regulation of establishment or maintenance of cell polarity
          regulation of intracellular steroid hormone receptor signaling pathway
          positive regulation of protein import into nucleus, translocation
          negative regulation of smooth muscle cell apoptotic process
          multicellular organism growth
          bone mineralization involved in bone maturation
          positive regulation of activated T cell proliferation
          regulation of cell proliferation
          positive regulation of tyrosine phosphorylation of Stat5 protein
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of DNA binding
          positive regulation of MAPK cascade
          protein kinase B signaling
          positive regulation of insulin-like growth factor receptor signaling pathway
          myoblast differentiation
          positive regulation of fat cell differentiation
          positive regulation of osteoblast differentiation
          positive regulation of glycogen biosynthetic process
          positive regulation of DNA replication
          positive regulation of glycolytic process
          positive regulation of mitotic nuclear division
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of glucose import
          positive regulation of Ras protein signal transduction
          insulin-like growth factor receptor signaling pathway
          phosphatidylinositol-mediated signaling
          positive regulation of fibroblast proliferation
          lung alveolus development
          cell development
          positive regulation of smooth muscle cell proliferation
          branching morphogenesis of an epithelial tube
          inner ear development
          chondroitin sulfate proteoglycan biosynthetic process
          positive regulation of epithelial cell proliferation
          positive regulation of peptidyl-tyrosine phosphorylation
          detection of mechanical stimulus involved in sensory perception
          regulation of protein metabolic process
          myoblast proliferation
          positive regulation of protein kinase B signaling
          regulation of calcium ion transport
          negative regulation of oocyte development
          lung vasculature development
          lung lobe morphogenesis
          Type I pneumocyte differentiation
          Type II pneumocyte differentiation
          prostate epithelial cord arborization involved in prostate glandular acinus morphogenesis
          prostate gland growth
          prostate gland epithelium morphogenesis
          prostate gland stromal morphogenesis
          negative regulation of androgen receptor signaling pathway
          ERK1 and ERK2 cascade
          negative regulation of ERK1 and ERK2 cascade
          positive regulation of calcineurin-NFAT signaling cascade
          positive regulation of protein serine/threonine kinase activity
          positive regulation of steroid hormone biosynthetic process
          negative regulation of release of cytochrome c from mitochondria
          extrinsic apoptotic signaling pathway in absence of ligand
          negative regulation of neuron death
          positive regulation of trophectodermal cell proliferation
          positive regulation of myoblast proliferation
          negative regulation of extrinsic apoptotic signaling pathway
          insulin receptor binding
          insulin-like growth factor receptor binding
          integrin binding
          hormone activity
          steroid binding
          protein binding
          growth factor activity
          protein serine/threonine kinase activator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0