Search
Search

ERCC6L2| Gene Symbol: ERCC6L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020207.4 | NP_064592.2 | 18 | 4059 | ||
| XM_011518641.2 | XP_011516943.1 | 17 | 3906 | |||
| XM_011518644.2 | XP_011516946.1 | 17 | 3851 | |||
| XM_011518645.2 | XP_011516947.1 | 18 | 4059 | |||
| XM_011518648.2 | XP_011516950.1 | 9 | 2131 | |||
| XM_011518650.2 | XP_011516952.1 | 18 | 3378 | |||
| XM_017014707.1 | XP_016870196.1 | 18 | 4046 | |||
| GenBank mRNA | AK308131.1 | - | 3 | 1094 | ||
| AL133663.1 | - | 13 | 2659 | |||
| BC022957.1 | - | 3 | 488 | 4995030 | ||
| BG108141.1 | - | 2 | 319 | |||