Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HSD3B1 SIRNA Assays ›
Gene Symbol:
Hsd3b1
Gene Name:
hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1
Gene Aliases:
-
Chromosome Location:
-
UniGene ID:
Rn.128814
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
286430
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Hsd3b1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001007719.3
NP_001007720.3
4
405
GenBank mRNA
BC086578.1
-
4
405
7306702
M38178.1
-
3
424
Back To Top
More Information
Panther Classification:
Molecular Function -
dehydratase
Gene Ontology Categories:
Function(s)
Process(es)
organic acid metabolic process
steroid biosynthetic process
progesterone biosynthetic process
estrogen biosynthetic process
steroid metabolic process
C21-steroid hormone metabolic process
male gonad development
female gonad development
response to herbicide
response to organic substance
response to metal ion
response to lithium ion
response to lead ion
response to organic cyclic compound
response to activity
diterpenoid metabolic process
insecticide metabolic process
biphenyl metabolic process
dibenzo-p-dioxin metabolic process
phenol-containing compound metabolic process
toluene metabolic process
hippocampus development
adrenal gland development
ovulation
granulocyte differentiation
response to nutrient levels
response to estradiol
cellular response to insulin stimulus
response to vitamin E
Leydig cell differentiation
response to L-ascorbic acid
response to drug
response to alkaloid
cellular response to fibroblast growth factor stimulus
response to ethanol
phosphatidylcholine metabolic process
response to arsenic-containing substance
response to cadmium ion
response to corticosterone
response to calcium ion
oxidation-reduction process
maternal process involved in female pregnancy
response to fungicide
testosterone biosynthetic process
cellular response to hydrogen peroxide
response to fatty acid
cellular response to antibiotic
cellular response to mercury ion
cellular response to cAMP
cellular response to glucose stimulus
cellular response to gonadotropin stimulus
cellular response to follicle-stimulating hormone stimulus
cellular response to luteinizing hormone stimulus
cellular response to methylmercury
cellular response to dexamethasone stimulus
cellular response to transforming growth factor beta stimulus
3-beta-hydroxy-delta5-steroid dehydrogenase activity
steroid delta-isomerase activity
steroid binding
dihydrotestosterone 17-beta-dehydrogenase activity
NAD binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0