Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HBB SIRNA Assays ›
Gene Symbol:
Hbb
Gene Name:
hemoglobin, beta
Gene Aliases:
-
Chromosome Location:
Chr.1: 168971269 - 168972680 on Build Rnor_6.0
UniGene ID:
Rn.202945
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
286807
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Hbb
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_033234.1
NP_150237.1
1
109
GenBank mRNA
BC058448.1
-
1
118
6921704
FQ209466.1
-
1
114
FQ209544.1
-
1
114
FQ209725.1
-
1
116
FQ209747.1
-
1
114
FQ209754.1
-
1
114
FQ209760.1
-
1
114
FQ209789.1
-
1
114
FQ209796.1
-
1
114
FQ209806.1
-
1
116
FQ209822.1
-
114
FQ209826.1
-
1
123
FQ209841.1
-
1
116
FQ209870.1
-
1
116
FQ209871.1
-
1
114
FQ209903.1
-
1
114
FQ209929.1
-
1
114
FQ209937.1
-
1
114
FQ209954.1
-
1
114
FQ209961.1
-
1
113
FQ209989.1
-
1
114
FQ209996.1
-
1
114
FQ210038.1
-
1
116
FQ210054.1
-
1
114
FQ210058.1
-
1
114
FQ210090.1
-
1
116
FQ210123.1
-
1
114
FQ210127.1
-
1
114
FQ210137.1
-
1
114
FQ210175.1
-
1
112
FQ210181.1
-
1
116
FQ210211.1
-
1
114
FQ210216.1
-
1
113
FQ210218.1
-
1
114
FQ210280.1
-
1
137
FQ210348.1
-
1
114
FQ210353.1
-
1
114
FQ210360.1
-
1
114
FQ210373.1
-
1
114
FQ210415.1
-
1
114
FQ210449.1
-
1
114
FQ210466.1
-
1
114
FQ210482.1
-
1
114
FQ210486.1
-
1
116
FQ210495.1
-
1
114
FQ210497.1
-
1
115
FQ210499.1
-
1
114
FQ210511.1
-
1
114
FQ210523.1
-
1
114
FQ210534.1
-
1
114
FQ210558.1
-
1
114
FQ210602.1
-
1
114
FQ210676.1
-
1
114
FQ210677.1
-
1
114
FQ210735.1
-
1
113
FQ210771.1
-
1
114
FQ210783.1
-
1
106
FQ210789.1
-
1
114
FQ210794.1
-
1
114
FQ210798.1
-
1
114
FQ210806.1
-
1
114
FQ210826.1
-
1
114
FQ210828.1
-
1
111
FQ210829.1
-
1
114
FQ210838.1
-
1
114
FQ210852.1
-
1
114
FQ210875.1
-
1
114
FQ210912.1
-
1
114
FQ210937.1
-
1
114
FQ210949.1
-
1
117
FQ210960.1
-
1
114
FQ210984.1
-
1
114
FQ210985.1
-
1
114
FQ211014.1
-
1
114
FQ211016.1
-
1
114
FQ211034.1
-
1
114
FQ211061.1
-
1
113
FQ211065.1
-
1
114
FQ211103.1
-
1
114
FQ211125.1
-
1
116
FQ211133.1
-
1
111
FQ211137.1
-
1
114
FQ211142.1
-
1
116
FQ211147.1
-
1
114
FQ211182.1
-
1
114
FQ211193.1
-
1
114
FQ211207.1
-
1
116
FQ211241.1
-
1
114
FQ211242.1
-
1
114
FQ211290.1
-
1
114
FQ211314.1
-
1
114
FQ211331.1
-
1
114
FQ211336.1
-
1
114
FQ211338.1
-
1
113
FQ211340.1
-
1
114
FQ211423.1
-
1
116
FQ211443.1
-
1
114
FQ211452.1
-
1
114
FQ211477.1
-
1
114
FQ211497.1
-
1
114
FQ211548.1
-
1
114
FQ211583.1
-
1
114
FQ211590.1
-
1
115
FQ211613.1
-
1
116
FQ211617.1
-
1
114
FQ211626.1
-
1
114
FQ211692.1
-
1
114
FQ211696.1
-
1
114
FQ211790.1
-
1
114
FQ211806.1
-
1
114
FQ211815.1
-
1
114
FQ211829.1
-
1
116
FQ211847.1
-
1
114
FQ211858.1
-
1
114
FQ211864.1
-
1
114
FQ211905.1
-
1
114
FQ211931.1
-
1
114
FQ211999.1
-
1
114
FQ212127.1
-
1
114
FQ212156.1
-
1
112
FQ212234.1
-
1
114
FQ212241.1
-
1
114
FQ212258.1
-
1
113
FQ212275.1
-
1
114
FQ212287.1
-
1
114
FQ212290.1
-
1
114
FQ212303.1
-
1
114
FQ212341.1
-
1
116
FQ212449.1
-
1
114
FQ213352.1
-
1
114
FQ213466.1
-
1
114
FQ213534.1
-
1
114
FQ213582.1
-
593
FQ213796.1
-
1
114
FQ214044.1
-
1
114
FQ214573.1
-
1
114
FQ217014.1
-
1
116
FQ217108.1
-
1
114
FQ217151.1
-
1
114
FQ217201.1
-
1
114
FQ217206.1
-
1
114
FQ217586.1
-
1
116
FQ217619.1
-
114
FQ217623.1
-
1
114
FQ217653.1
-
1
114
FQ217685.1
-
1
114
FQ217803.1
-
1
83
FQ218042.1
-
1
114
FQ218100.1
-
1
114
FQ218112.1
-
1
114
FQ218118.1
-
1
116
FQ218124.1
-
1
118
FQ218145.1
-
1
114
FQ218153.1
-
1
114
FQ218162.1
-
1
114
FQ218163.1
-
1
114
FQ218171.1
-
1
114
FQ218180.1
-
1
114
FQ218182.1
-
1
114
FQ218199.1
-
1
117
FQ218229.1
-
1
117
FQ218253.1
-
1
114
FQ218272.1
-
1
111
FQ218288.1
-
1
114
FQ218461.1
-
1
134
FQ218539.1
-
1
113
FQ218748.1
-
1
114
FQ218761.1
-
1
114
FQ218763.1
-
1
114
FQ218779.1
-
1
112
FQ219114.1
-
1
114
FQ219985.1
-
1
63
FQ220085.1
-
1
114
FQ220253.1
-
1
63
FQ221076.1
-
1
114
FQ221202.1
-
1
116
FQ221246.1
-
1
114
FQ221520.1
-
1
114
FQ221738.1
-
1
113
FQ221956.1
-
1
114
FQ221962.1
-
1
114
FQ222032.1
-
1
114
FQ222179.1
-
1
114
FQ222262.1
-
1
114
FQ222275.1
-
1
114
FQ222291.1
-
1
114
FQ222503.1
-
1
114
FQ222804.1
-
1
114
FQ222942.1
-
1
114
FQ222960.1
-
1
114
FQ223002.1
-
1
114
FQ223079.1
-
1
114
FQ223164.1
-
1
111
FQ223490.1
-
1
114
FQ223631.1
-
1
114
FQ223717.1
-
1
114
FQ223745.1
-
1
116
FQ223805.1
-
1
114
FQ223852.1
-
1
106
FQ223888.1
-
1
114
FQ223889.1
-
1
116
FQ223995.1
-
1
87
FQ224045.1
-
1
116
FQ224088.1
-
1
114
FQ224309.1
-
1
114
FQ224343.1
-
1
114
FQ224371.1
-
1
114
FQ224406.1
-
1
114
FQ224475.1
-
1
114
FQ224682.1
-
1
114
FQ224727.1
-
1
114
FQ224732.1
-
1
114
FQ225124.1
-
1
176
FQ225436.1
-
1
114
FQ225684.1
-
266
FQ225738.1
-
1
114
FQ225754.1
-
1
113
FQ225816.1
-
1
114
FQ225817.1
-
1
114
FQ226188.1
-
1
112
FQ226198.1
-
115
FQ226536.1
-
1
114
FQ226572.1
-
1
114
FQ226889.1
-
1
114
FQ227120.1
-
1
114
FQ227384.1
-
115
FQ227498.1
-
1
114
FQ227822.1
-
1
114
FQ227875.1
-
1
114
FQ228016.1
-
1
114
FQ228244.1
-
1
114
FQ228281.1
-
1
114
FQ228294.1
-
1
114
FQ228295.1
-
1
114
FQ228296.1
-
1
116
FQ228414.1
-
1
114
FQ228616.1
-
1
114
FQ228638.1
-
1
114
FQ229015.1
-
1
114
FQ229033.1
-
1
114
FQ229489.1
-
1
114
FQ230471.1
-
1
114
FQ230486.1
-
1
112
FQ230503.1
-
1
114
FQ230508.1
-
1
114
FQ230541.1
-
1
114
FQ230861.1
-
1
114
FQ230866.1
-
1
114
FQ230889.1
-
1
114
FQ231191.1
-
1
113
FQ231199.1
-
1
114
FQ231208.1
-
1
113
FQ231230.1
-
1
114
FQ231253.1
-
1
114
FQ231284.1
-
1
116
FQ231286.1
-
1
114
FQ232390.1
-
382
FQ232655.1
-
1
146
FQ233311.1
-
1
189
FQ234713.1
-
1
116
FQ234942.1
-
1
112
FQ235254.1
-
750
M17084.1
-
1
66
X16417.1
-
1
109
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
glutathione metabolic process
regulation of eIF2 alpha phosphorylation by heme
oxygen transport
hemopoiesis
erythrocyte development
protein heterooligomerization
oxygen transporter activity
iron ion binding
oxygen binding
heme binding
hemoglobin alpha binding
hemoglobin beta binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-sk2zd:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0