Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other H2-K1 SIRNA Assays ›
          Gene Symbol:
          H2-K1 H2-D1
          Gene Name:
          histocompatibility 2, K1, K region
          histocompatibility 2, D region locus 1
          Gene Aliases:
          H-2K, H-2K(d), H2-K, K-f
          H-2D, H2-D
          Chromosome Location:
          Chr.17: 33996012 - 34000342 on Build GRCm38
          Chromosome Location:
          Chr.17: 35263094 - 35267497 on Build GRCm38
          UniGene ID:
          Mm.422886
          Mm.439675
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 287394

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: H2-K1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001001892.2 NP_001001892.2 8 1215
          GenBank mRNA AK150790.1 - 6 1449
          AK150967.1 - 8 1166
          AK153419.1 - 8 1216
          AK159321.1 - 8 1166
          AK160102.1 - 8 1167
          AK168517.1 - 8 1140
          BC011306.1 - 7 889 3156482
          BC054430.1 - 7 1087
          BC080756.1 - 2 1183 4947189
          BC151193.1 - 8 1174 9088081
          DQ346646.1 - 8 1136
          DQ346647.1 - 8 1136
          DQ346648.1 - 8 1136
          J00395.1 - 5 524
          J00400.1 - 7 881
          J00402.1 - 8 1161
          K01762.1 - 5 338
          L23494.1 - 8 1144
          L23495.1 - 8 1171
          L36065.1 - 8 1155
          L36306.1 - 8 1155
          L36307.1 - 8 1155
          L36308.1 - 8 1155
          L36309.1 - 8 1155
          L36310.1 - 8 1155
          L36311.1 - 8 1155
          L36312.1 - 8 1155
          M11975.1 - 8 1076
          M13197.1 - 1 29
          M14827.1 - 7 1016
          M23894.1 - 4 254
          M23895.1 - 4 254
          M23896.1 - 4 254
          M23899.1 - 3 115
          M23900.1 - 2 93
          M23901.1 - 1 31
          M23902.1 - 2 79
          M58156.1 - 8 1151
          M69071.1 - 8 1136
          M69072.1 - 8 1136
          M69073.1 - 8 1136
          S70184.1 - 6 825
          U47328.1 - 8 1139
          U47329.1 - 8 1136
          U47330.1 - 8 1112
          X00172.1 - 5 365
          Gene Symbol: H2-D1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA J00406.1 - 3 114
          M23445.1 - 161
          M69069.1 - 1072
          U47326.1 - 1127
          X00246.1 - 6 887

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of T cell mediated cytotoxicity
          immune system process
          antigen processing and presentation of peptide antigen via MHC class I
          antigen processing and presentation of endogenous peptide antigen via MHC class I via ER pathway, TAP-dependent
          immune response
          antigen processing and presentation
          antigen processing and presentation of exogenous peptide antigen via MHC class I
          defense response to bacterium
          inner ear development
          receptor binding
          protein binding
          beta-2-microglobulin binding
          peptide binding
          peptide antigen binding
          T cell receptor binding
          poly(A) RNA binding
          TAP binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0