Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TPM1 SIRNA Assays ›
Gene Symbol:
TPM1
Gene Name:
tropomyosin 1 (alpha)
Gene Aliases:
C15orf13, CMD1Y, CMH3, HEL-S-265, HTM-alpha, LVNC9, TMSA
Chromosome Location:
Chr.15: 63042196 - 63071915 on Build GRCh38
UniGene ID:
Hs.133892
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
288546
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TPM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000366.5
NP_000357.3
4
631
NM_001018004.1
NP_001018004.1
4
631
NM_001018005.1
NP_001018005.1
4
631
NM_001018006.1
NP_001018006.1
4
631
NM_001018007.1
NP_001018007.1
4
631
NM_001018008.1
NP_001018008.1
3
471
NM_001018020.1
NP_001018020.1
4
631
NM_001301244.1
NP_001288173.1
4
631
NM_001301289.1
NP_001288218.1
3
471
XM_005254637.1
XP_005254694.1
5
712
XM_005254638.3
XP_005254695.2
4
1074
XM_005254639.3
XP_005254696.2
4
1074
XM_005254645.2
XP_005254702.2
4
1074
XM_005254646.2
XP_005254703.1
3
457
XM_005254647.3
XP_005254704.1
3
457
XM_005254650.3
XP_005254707.1
3
457
XM_005254651.1
XP_005254708.1
3
457
XM_005254652.1
XP_005254709.1
3
457
XM_005254653.2
XP_005254710.1
3
457
XM_006720667.3
XP_006720730.2
4
1074
XM_017022534.1
XP_016878023.1
4
1074
XM_017022535.1
XP_016878024.1
4
586
XM_017022536.1
XP_016878025.1
4
1074
XM_017022537.1
XP_016878026.1
4
586
XM_017022538.1
XP_016878027.1
4
1074
XM_017022539.1
XP_016878028.1
4
1074
XM_017022540.1
XP_016878029.1
3
457
XM_017022541.1
XP_016878030.1
3
457
GenBank mRNA
AB209041.1
-
3
508
AF474156.1
-
3
233
AK057989.1
-
6
417
AK092051.1
-
3
1348
AK092573.1
-
2
1453
AK131384.1
-
5
645
AK298625.1
-
3
347
AK299387.1
-
4
520
AK301319.1
-
3
367
AK309170.1
-
4
840
AL050179.1
-
4
512
AY640414.1
-
4
440
AY640415.1
-
4
440
BC007433.2
-
4
581
2962925
BC050473.1
-
3
458
BC053545.1
-
3
469
5730451
BG107949.1
-
4
638
BX458841.2
-
4
500
CN423344.1
-
4
569
DQ424903.1
-
3
332
GQ472198.1
-
4
581
KU178366.1
-
4
440
KU178367.1
-
4
440
M19713.1
-
4
496
M19714.1
-
4
382
M19715.1
-
3
277
X12369.1
-
3
270
Z24727.1
-
655
Back To Top
More Information
Panther Classification:
Molecular Function -
actin binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
positive regulation of heart rate by epinephrine
movement of cell or subcellular component
muscle contraction
regulation of muscle contraction
cytoskeleton organization
regulation of heart contraction
regulation of cell shape
muscle filament sliding
negative regulation of cell migration
ruffle organization
positive regulation of ATPase activity
cellular response to reactive oxygen species
wound healing
sarcomere organization
positive regulation of cell adhesion
positive regulation of stress fiber assembly
ventricular cardiac muscle tissue morphogenesis
cardiac muscle contraction
negative regulation of vascular smooth muscle cell proliferation
negative regulation of vascular associated smooth muscle cell migration
actin binding
structural constituent of cytoskeleton
protein binding
cytoskeletal protein binding
structural constituent of muscle
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0