Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TSPAN4 SIRNA Assays ›
Gene Symbol:
TSPAN4
Gene Name:
tetraspanin 4
Gene Aliases:
NAG-2, NAG2, TETRASPAN, TM4SF7, TSPAN-4
Chromosome Location:
Chr.11: 842824 - 867116 on Build GRCh38
UniGene ID:
Hs.654836
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
288560
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TSPAN4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001025234.1
NP_001020405.1
5, 6
552
NM_001025235.1
NP_001020406.1
5, 6
551
NM_001025236.1
NP_001020407.1
5, 6
556
NM_001025237.1
NP_001020408.1
5, 6
588
NM_001025238.1
NP_001020409.1
4, 5
438
NM_001025239.1
NP_001020410.1
4, 5
458
NM_003271.4
NP_003262.1
5, 6
538
XM_005253102.1
XP_005253159.1
4, 5
457
XM_005253104.1
XP_005253161.1
3, 4
266
XM_011520336.1
XP_011518638.1
4, 5
897
XM_011520337.2
XP_011518639.1
4, 5
759
XM_011520338.1
XP_011518640.1
4, 5
479
XM_011520339.2
XP_011518641.1
4, 5
654
XM_011520340.2
XP_011518642.1
4, 5
537
XM_011520341.2
XP_011518643.1
4, 5
458
GenBank mRNA
AF022813.1
-
4, 5
433
AF054841.1
-
4, 5
447
AK054788.1
-
1, 2
1239
AK095212.1
-
3, 4
1910
AK291089.1
-
5, 6
513
BC000389.2
-
5, 6
464
2821236
BC019314.2
-
4, 5
449
2821236
BC093031.1
-
5, 6
507
30334510
BG752017.1
-
5, 6
545
BI820547.1
-
5, 6
608
BQ679002.1
-
558
BT006776.1
-
3, 4
329
CR457108.1
-
3, 4
329
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein complex assembly
cell surface receptor signaling pathway
antigen binding
integrin binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0