Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other APOA4 SIRNA Assays ›
          Gene Symbol:
          APOA4
          Gene Name:
          apolipoprotein A4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 116820702 - 116823295 on Build GRCh38
          UniGene ID:
          Hs.1247
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 289146

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: APOA4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000482.3 NP_000473.2 3 1159
          GenBank mRNA BC074764.2 - 3 1093 30915177
          BC113594.1 - 3 1084 8322646
          BC113596.1 - 3 1084 8322648
          M10373.1 - 1 653
          M13654.1 - 3 1106
          M14566.1 - 2 995
          X13629.1 - 3 1100

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          apolipoprotein

          Gene Ontology Categories:

          Function(s) Process(es)

          retinoid metabolic process
          innate immune response in mucosa
          cholesterol biosynthetic process
          lipid transport
          response to lipid hydroperoxide
          leukocyte cell-cell adhesion
          cholesterol metabolic process
          positive regulation of cholesterol esterification
          positive regulation of triglyceride catabolic process
          removal of superoxide radicals
          regulation of intestinal cholesterol absorption
          neuron projection regeneration
          regulation of cholesterol transport
          cholesterol efflux
          phospholipid efflux
          chylomicron remodeling
          very-low-density lipoprotein particle remodeling
          high-density lipoprotein particle remodeling
          chylomicron assembly
          high-density lipoprotein particle assembly
          negative regulation of plasma lipoprotein particle oxidation
          response to stilbenoid
          lipoprotein metabolic process
          lipoprotein biosynthetic process
          cholesterol homeostasis
          hydrogen peroxide catabolic process
          reverse cholesterol transport
          multicellular organism lipid catabolic process
          cellular protein metabolic process
          positive regulation of fatty acid biosynthetic process
          phosphatidylcholine metabolic process
          positive regulation of lipoprotein lipase activity
          lipid homeostasis
          protein-lipid complex assembly
          triglyceride homeostasis
          lipid transporter activity
          copper ion binding
          protein binding
          lipid binding
          cholesterol binding
          antioxidant activity
          cholesterol transporter activity
          phosphatidylcholine binding
          protein homodimerization activity
          phosphatidylcholine-sterol O-acyltransferase activator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0