Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002506.2 | NP_002497.2 | 3 | 290 | ||
XM_006710663.3 | XP_006710726.1 | 2 | 183 | |||
XM_011541518.2 | XP_011539820.1 | 3 | 3328 | |||
GenBank mRNA | AF411526.1 | - | 2 | 194 | ||
BC032517.2 | - | 3 | 295 | |||
BC126148.1 | - | 2 | 217 | 8991864 | ||
BC126150.1 | - | 2 | 217 | 8991866 | ||
BT019733.1 | - | 1 | 121 | |||
CR541855.1 | - | 1 | 121 | |||
X52599.1 | - | 3 | 290 |