Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HMGA2 SIRNA Assays ›
          Gene Symbol:
          HMGA2
          Gene Name:
          high mobility group AT-hook 2
          Gene Aliases:
          BABL, HMGI-C, HMGIC, LIPO, STQTL9
          Chromosome Location:
          Chr.12: 65824460 - 65966291 on Build GRCh38
          UniGene ID:
          Hs.505924
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 289435

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HMGA2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001300918.1 NP_001287847.1 3 1039
          NM_001300919.1 NP_001287848.1 3 1039
          NM_003483.4 NP_003474.1 3 1039
          NM_003484.1 NP_003475.1 3 1039
          XM_017019989.1 XP_016875478.1 3 688
          GenBank mRNA AB209853.1 - 3 601
          AF533651.1 - 2 103
          AF533652.1 - 2 137
          AF533653.1 - 131
          AF533654.1 - 2 131
          AI290477.1 - 2 117
          AY601861.1 - 3 1039
          AY601862.1 - 3 1039
          AY601863.1 - 3 1039
          AY601864.1 - 3 1039
          AY601865.1 - 3 1039
          AY601866.1 - 3 1039
          AY601867.1 - 3 1039
          U28749.1 - 3 1039
          X92518.1 - 3 1538
          Z31595.1 - 3 1040

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_032070, U89695.
          This siRNA also targets the following transcripts in Mouse: NM_010441, BC052158, BC085085, BC069985, AK028346, and others.

          Panther Classification:

          Molecular Function -

          endodeoxyribonuclease

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          epithelial to mesenchymal transition
          chondrocyte differentiation
          mesodermal-endodermal cell signaling
          base-excision repair
          chromatin organization
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          mitotic nuclear division
          mitotic G2 DNA damage checkpoint
          multicellular organism development
          response to virus
          regulation of cell cycle process
          positive regulation of gene expression
          chromosome condensation
          chromosome breakage
          heterochromatin assembly
          histone H2A-S139 phosphorylation
          senescence-associated heterochromatin focus assembly
          endodermal cell differentiation
          chondrocyte proliferation
          regulation of growth
          DNA damage response, detection of DNA damage
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          negative regulation of DNA binding
          negative regulation by host of viral transcription
          fat cell differentiation
          negative regulation of single stranded viral RNA replication via double stranded DNA intermediate
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          mesodermal cell differentiation
          mesenchymal cell differentiation
          stem cell differentiation
          cell division
          positive regulation of cell cycle arrest
          oncogene-induced cell senescence
          regulation of stem cell population maintenance
          positive regulation of stem cell proliferation
          positive regulation of transcription regulatory region DNA binding
          positive regulation of cellular response to X-ray
          positive regulation of cellular senescence
          positive regulation of response to DNA damage stimulus
          negative regulation of double-strand break repair via nonhomologous end joining
          regulation of cellular response to drug
          regulatory region DNA binding
          transcription factor activity, transcription factor binding
          core promoter binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          AT DNA binding
          DNA-(apurinic or apyrimidinic site) lyase activity
          DNA-dependent protein kinase activity
          protein binding
          transcription factor binding
          DNA binding, bending
          nucleosomal DNA binding
          cAMP response element binding
          MH2 domain binding
          MH1 domain binding
          SMAD binding
          5'-deoxyribose-5-phosphate lyase activity
          C2H2 zinc finger domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-kjp6x:80/100.66.78.163:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0