Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MBNL1 SIRNA Assays ›
Gene Symbol:
MBNL1
Gene Name:
muscleblind like splicing regulator 1
Gene Aliases:
EXP, MBNL
Chromosome Location:
Chr.3: 152243656 - 152465780 on Build GRCh38
UniGene ID:
Hs.201858
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
28944
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MBNL1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001314057.1
NP_001300986.1
6
1223
NM_021038.4
NP_066368.2
5
2008
NM_207292.2
NP_997175.1
5
2008
NM_207293.1
NP_997176.1
4
1383
NM_207294.1
NP_997177.1
3
1179
NM_207295.1
NP_997178.1
3
1179
NM_207296.1
NP_997179.1
4
1383
NM_207297.1
NP_997180.1
4
1383
XM_005247457.4
XP_005247514.1
6
1576
XM_005247458.4
XP_005247515.1
6
2830
XM_005247460.4
XP_005247517.1
6
1539
XM_005247461.4
XP_005247518.1
6
1576
XM_005247462.4
XP_005247519.1
6
1576
XM_005247463.4
XP_005247520.1
6
1576
XM_005247464.4
XP_005247521.1
5
1634
XM_005247465.4
XP_005247522.1
6
1576
XM_005247466.4
XP_005247523.1
5
1611
XM_005247467.4
XP_005247524.1
6
1576
XM_005247468.4
XP_005247525.1
6
1576
XM_005247471.4
XP_005247528.1
5
1606
XM_005247474.3
XP_005247531.1
4
494
XM_005247475.3
XP_005247532.1
5
817
XM_005247476.4
XP_005247533.1
4
584
XM_005247477.4
XP_005247534.1
4
583
XM_006713639.1
XP_006713702.1
6
1558
XM_011512843.2
XP_011511145.1
5
1372
XM_011512845.2
XP_011511147.1
5
1372
XM_011512846.2
XP_011511148.1
5
1372
XM_011512847.2
XP_011511149.1
4
1335
XM_011512848.2
XP_011511150.1
6
1180
XM_011512849.2
XP_011511151.1
4
1279
XM_017006423.1
XP_016861912.1
5
1827
XM_017006424.1
XP_016861913.1
5
1654
XM_017006425.1
XP_016861914.1
5
1656
XM_017006426.1
XP_016861915.1
5
1654
XM_017006427.1
XP_016861916.1
5
1801
XM_017006428.1
XP_016861917.1
5
1465
XM_017006429.1
XP_016861918.1
5
1548
XM_017006430.1
XP_016861919.1
5
1612
XM_017006431.1
XP_016861920.1
5
1689
XM_017006432.1
XP_016861921.1
5
1611
XM_017006433.1
XP_016861922.1
5
1611
XM_017006434.1
XP_016861923.1
5
1617
XM_017006435.1
XP_016861924.1
5
624
XM_017006436.1
XP_016861925.1
5
2737
XM_017006437.1
XP_016861926.1
5
1792
XM_017006438.1
XP_016861927.1
4
1279
XM_017006439.1
XP_016861928.1
5
817
XM_017006440.1
XP_016861929.1
4
551
XM_017006441.1
XP_016861930.1
4
584
XM_017006442.1
XP_016861931.1
4
687
XM_017006443.1
XP_016861932.1
4
695
GenBank mRNA
AB007888.1
-
5
2008
AF255334.1
-
3
901
AF395876.1
-
3
390
AF401998.1
-
4
594
AF497718.1
-
4
594
AF497719.1
-
4
594
AJ308400.1
-
4
623
AK297133.1
-
2
99
AK304089.1
-
3
303
AK307420.1
-
3
303
BC043493.1
-
5
1513
6044671
BC050535.1
-
6
744
6042311
Y13829.1
-
4
1105
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
mRNA processing
nervous system development
RNA splicing
embryonic limb morphogenesis
regulation of RNA splicing
myoblast differentiation
regulatory region RNA binding
RNA binding
double-stranded RNA binding
protein binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0