Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LIMS1 SIRNA Assays ›
          Gene Symbol:
          LIMS1
          Gene Name:
          LIM zinc finger domain containing 1
          Gene Aliases:
          PINCH, PINCH-1, PINCH1
          Chromosome Location:
          Chr.2: 108534355 - 108687246 on Build GRCh38
          UniGene ID:
          Hs.597715
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 289501

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LIMS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001193482.1 NP_001180411.1 10 1161
          NM_001193483.2 NP_001180412.1 10 1306
          NM_001193484.1 NP_001180413.1 10 1282
          NM_001193485.2 NP_001180414.1 10 1511
          NM_001193488.1 NP_001180417.1 10 1229
          NM_004987.5 NP_004978.2 10 1191
          XM_005263946.1 XP_005264003.1 11 1503
          XM_005263948.3 XP_005264005.1 11 1556
          XM_005263949.1 XP_005264006.1 11 1498
          XM_005263950.1 XP_005264007.1 11 1339
          XM_005263951.4 XP_005264008.1 11 1775
          XM_017004091.1 XP_016859580.1 11 1525
          XM_017004092.1 XP_016859581.1 11 1520
          XM_017004093.1 XP_016859582.1 11 1569
          XM_017004094.1 XP_016859583.1 11 1804
          GenBank mRNA AF527759.1 - 10 1511
          AF527760.1 - 10 1263
          AF527761.1 - 10 1239
          AF527762.1 - 10 1172
          AF527763.1 - 10 1161
          AK296992.1 - 10 1261
          AK302411.1 - 10 1286
          AK304260.1 - 10 1282
          BC005341.1 - 10 1229 3838632
          U09284.2 - 10 1181

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cell junction protein

          Gene Ontology Categories:

          Function(s) Process(es)

          epithelial to mesenchymal transition
          cell aging
          positive regulation of gene expression
          positive regulation of cell-substrate adhesion
          cell junction assembly
          positive regulation of GTPase activity
          establishment of protein localization
          negative regulation of transcription, DNA-templated
          regulation of epithelial cell proliferation
          positive regulation of focal adhesion assembly
          cellular response to transforming growth factor beta stimulus
          positive regulation of substrate adhesion-dependent cell spreading
          protein binding
          zinc ion binding
          protein kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0