Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KIR3DS1 SIRNA Assays ›
          Gene Symbol:
          KIR3DS1 KIR3DL1
          Gene Name:
          killer cell immunoglobulin like receptor, three Ig domains and short cytoplasmic tail 1
          killer cell immunoglobulin like receptor, three Ig domains and long cytoplasmic tail 1
          Gene Aliases:
          CD158E2, KIR-123FM, KIR-G1, NKAT-10, NKAT10
          CD158E1, KIR, KIR3DL1/S1, KIR3DL2, NKAT-3, NKAT3, NKB1, NKB1B
          Chromosome Location:
          -
          Chromosome Location:
          Chr.19: 54816438 - 54830778 on Build GRCh38
          UniGene ID:
          Hs.743496
          Hs.645228
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 289674

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: KIR3DS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001083539.2 NP_001077008.1 3 198
          GenBank mRNA AF022044.1 - 3 174
          AJ417558.3 - 1 110
          AK301140.1 - 3 198
          BC098288.1 - 3 172 40026980
          BC105679.1 - 3 172 40004313
          DC390157.1 - 3 198
          EF370406.1 - 2 122
          L76661.1 - 3 172
          U73396.1 - 3 161
          X97233.1 - 3 161
          Gene Symbol: KIR3DL1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001322168.1 NP_001309097.1 3 224
          NM_013289.2 NP_037421.2 3 224
          GenBank mRNA AF022049.1 - 3 161
          AF103010.1 - 3 161
          AF103011.1 - 3 161
          AF103012.1 - 3 161
          AF103013.1 - 3 161
          AF208688.1 - 1 98
          AF208689.1 - 1 98
          AF262968.1 - 1 80
          AF262971.1 - 1 80
          AF262972.1 - 1 80
          AF262973.1 - 3 194
          AJ417556.1 - 1 110
          AJ417557.1 - 1 110
          AY366236.1 - 3 199
          AY366249.1 - 3 161
          AY366250.1 - 3 187
          AY366251.1 - 3 161
          AY366252.1 - 3 161
          AY760050.1 - 3 161
          AY789065.1 - 3 176
          AY789066.1 - 3 161
          AY789072.1 - 3 161
          BC028206.1 - 3 188 5215674
          BC101540.1 - 3 180 8069046
          BC101542.1 - 3 180 8069048
          EF462885.1 - 1 91
          EF472674.1 - 1 91
          EF472675.1 - 1 91
          EF472676.1 - 1 91
          EF472677.1 - 1 91
          EF472678.1 - 1 91
          EF472679.1 - 1 91
          L41269.1 - 3 180
          U30273.1 - 3 163
          U30274.1 - 3 224
          U31416.1 - 3 186
          U33328.1 - 3 186
          X94262.1 - 3 162

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          immune response
          signal transduction
          natural killer cell activation
          innate immune response
          natural killer cell mediated cytotoxicity
          regulation of immune response
          MHC class I receptor activity
          HLA-B specific inhibitory MHC class I receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0