Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PPARA SIRNA Assays ›
          Gene Symbol:
          PPARA
          Gene Name:
          peroxisome proliferator activated receptor alpha
          Gene Aliases:
          NR1C1, PPAR, PPARalpha, hPPAR
          Chromosome Location:
          Chr.22: 46150534 - 46243756 on Build GRCh38
          UniGene ID:
          Hs.103110
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 289995

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PPARA
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001001928.2 NP_001001928.1 5 576
          NM_005036.4 NP_005027.2 6 660
          XM_005261655.3 XP_005261712.1 6 953
          XM_005261656.3 XP_005261713.1 7 1117
          XM_006724269.3 XP_006724332.1 6 719
          XM_006724270.3 XP_006724333.1 5 625
          XM_011530239.2 XP_011528541.1 7 809
          XM_011530240.2 XP_011528542.1 7 812
          XM_011530241.2 XP_011528543.1 6 717
          XM_011530242.2 XP_011528544.1 5 539
          XM_011530243.2 XP_011528545.1 4 780
          XM_011530244.2 XP_011528546.1 7 812
          XM_011530245.2 XP_011528547.1 5 628
          XM_017028839.1 XP_016884328.1 8 987
          XM_017028840.1 XP_016884329.1 7 812
          GenBank mRNA AB307690.1 - 3 394
          AK289821.1 - 6 953
          BC000052.2 - 5 604
          CR456547.1 - 4 530
          CR457435.1 - 3 394
          EU395809.1 - 3 394
          EU650667.1 - 3 394
          HQ692862.1 - 3 394
          L02932.1 - 5 610
          S74349.1 - 4 517
          Y07619.1 - 3 394

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          C4 zinc finger nuclear receptor DNA-binding transcription factor zinc finger transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          response to hypoxia
          transcription initiation from RNA polymerase II promoter
          lipid metabolic process
          fatty acid metabolic process
          heart development
          epidermis development
          negative regulation of macrophage derived foam cell differentiation
          negative regulation of receptor biosynthetic process
          negative regulation of cholesterol storage
          negative regulation of sequestering of triglyceride
          fatty acid transport
          intracellular receptor signaling pathway
          positive regulation of fatty acid beta-oxidation
          negative regulation of protein binding
          negative regulation of appetite
          response to insulin
          circadian regulation of gene expression
          behavioral response to nicotine
          wound healing
          lipoprotein metabolic process
          regulation of circadian rhythm
          steroid hormone mediated signaling pathway
          cellular lipid metabolic process
          positive regulation of gluconeogenesis
          negative regulation of blood pressure
          negative regulation of glycolytic process
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of fatty acid oxidation
          negative regulation of inflammatory response
          enamel mineralization
          regulation of glycolytic process by positive regulation of transcription from RNA polymerase II promoter
          regulation of cellular ketone metabolic process by positive regulation of transcription from RNA polymerase II promoter
          regulation of lipid transport by positive regulation of transcription from RNA polymerase II promoter
          negative regulation of neuron death
          negative regulation of pri-miRNA transcription from RNA polymerase II promoter
          negative regulation of leukocyte cell-cell adhesion
          negative regulation of transcription regulatory region DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II repressing transcription factor binding
          transcriptional activator activity, RNA polymerase II transcription factor binding
          transcription coactivator binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          steroid hormone receptor activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          protein binding
          transcription factor binding
          drug binding
          zinc ion binding
          lipid binding
          phosphatase binding
          protein domain specific binding
          ubiquitin conjugating enzyme binding
          protein complex binding
          sequence-specific DNA binding
          NFAT protein binding
          MDM2/MDM4 family protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0