Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ADAM22 SIRNA Assays ›
Gene Symbol:
ADAM22
Gene Name:
ADAM metallopeptidase domain 22
Gene Aliases:
ADAM 22, MDC2
Chromosome Location:
Chr.7: 87934143 - 88202889 on Build GRCh38
UniGene ID:
Hs.256398
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
29514
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ADAM22
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001324417.1
NP_001311346.1
22
2216
NM_001324418.1
NP_001311347.1
22
2219
NM_001324419.1
NP_001311348.1
22
2216
NM_001324420.1
NP_001311349.1
22
2219
NM_001324421.1
NP_001311350.1
22
2219
NM_004194.4
NP_004185.1
22
2219
NM_016351.5
NP_057435.2
22
2219
NM_021721.4
NP_068367.1
22
2219
NM_021722.5
NP_068368.2
22
2219
NM_021723.4
NP_068369.1
22
2219
XM_005250445.3
XP_005250502.1
21
2067
XM_006716028.3
XP_006716091.1
21
2067
XM_006716029.3
XP_006716092.1
21
2067
XM_011516318.1
XP_011514620.1
22
2111
XM_011516319.2
XP_011514621.1
22
2195
XM_011516320.2
XP_011514622.1
21
2067
XM_011516321.2
XP_011514623.1
21
2067
XM_011516322.1
XP_011514624.1
22
2111
XM_011516323.1
XP_011514625.1
22
2111
XM_011516324.1
XP_011514626.1
22
1783
XM_017012329.1
XP_016867818.1
23
2162
XM_017012330.1
XP_016867819.1
23
2162
XM_017012331.1
XP_016867820.1
22
2192
XM_017012332.1
XP_016867821.1
22
2111
XM_017012333.1
XP_016867822.1
22
2111
XM_017012334.1
XP_016867823.1
23
2162
XM_017012335.1
XP_016867824.1
23
2162
XM_017012336.1
XP_016867825.1
22
2111
XM_017012337.1
XP_016867826.1
23
2162
XM_017012338.1
XP_016867827.1
22
2190
XM_017012339.1
XP_016867828.1
23
2162
XM_017012340.1
XP_016867829.1
11
842
XM_017012341.1
XP_016867830.1
21
1839
GenBank mRNA
AB009671.1
-
22
2017
AF073291.2
-
22
1942
AF155381.1
-
22
1942
AF155382.1
-
22
1942
AF158637.1
-
21
1779
AK125432.1
-
21
2082
AK291010.1
-
22
2066
BC125118.1
-
22
1973
40118904
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001007220, NM_001098225, NM_001007221, HM004095, AB009674, and others.
Panther Classification:
Molecular Function -
metalloprotease
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
cell adhesion
negative regulation of cell adhesion
central nervous system development
adult locomotory behavior
myelination in peripheral nervous system
metalloendopeptidase activity
integrin binding
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline