Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EFCAB6 SIRNA Assays ›
Gene Symbol:
EFCAB6
Gene Name:
EF-hand calcium binding domain 6
Gene Aliases:
DJBP, HSCBCIP1, dJ185D5.1
Chromosome Location:
Chr.22: 43528549 - 43812403 on Build GRCh38
UniGene ID:
Hs.658996
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
30203
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
286.00
Your Price
Online offer:
277.65
277.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EFCAB6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022785.3
NP_073622.2
23, 24
3198
NM_198856.2
NP_942153.1
21, 22
2840
XM_005261704.2
XP_005261761.1
23, 24
3138
XM_005261705.3
XP_005261762.1
22, 23
3052
XM_011530316.1
XP_011528618.1
24, 25
3425
XM_011530317.2
XP_011528619.1
22, 23
2985
XM_011530318.2
XP_011528620.1
22, 23
3081
XM_011530319.2
XP_011528621.1
21, 22
2773
XM_011530320.2
XP_011528622.1
24, 25
3237
XM_011530321.2
XP_011528623.1
21, 22
2768
XM_011530322.2
XP_011528624.1
22, 23
2896
XM_011530323.2
XP_011528625.1
19, 20
2554
XM_011530325.2
XP_011528627.1
17, 18
2299
XM_011530326.2
XP_011528628.1
24, 25
3237
XM_011530327.2
XP_011528629.1
14, 15
2046
XM_011530329.1
XP_011528631.1
10, 11
1518
XM_017028910.1
XP_016884399.1
24, 25
3237
XM_017028911.1
XP_016884400.1
24, 25
3237
XM_017028912.1
XP_016884401.1
20, 21
2896
GenBank mRNA
AB051459.1
-
9, 10
1381
AB073862.1
-
3, 4
392
AK027241.1
-
3, 4
396
AK225596.1
-
3, 4
397
AK225605.1
-
3, 4
397
AK292411.1
-
23, 24
3138
AK292498.1
-
21, 22
2817
AK309013.1
-
4, 5
596
BC039315.1
-
21, 22
2693
5297436
Back To Top
More Information
Panther Classification:
Molecular Function -
calmodulin-related
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
calcium ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-9rjdb:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline