Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCDC14 SIRNA Assays ›
Gene Symbol:
CCDC14
Gene Name:
coiled-coil domain containing 14
Gene Aliases:
-
Chromosome Location:
Chr.3: 123887383 - 123961412 on Build GRCh38
UniGene ID:
Hs.645028
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
30272
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CCDC14
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308317.1
NP_001295246.1
6, 7
862
NM_022757.4
NP_073594.4
6, 7
795
XM_005247710.4
XP_005247767.1
7, 8
918
XM_005247711.4
XP_005247768.1
7, 8
920
XM_005247713.2
XP_005247770.1
4, 5
692
XM_005247714.2
XP_005247771.1
3, 4
571
XM_005247715.4
XP_005247772.1
7, 8
921
XM_005247716.4
XP_005247773.1
7, 8
921
XM_006713731.3
XP_006713794.1
7, 8
920
XM_006713733.3
XP_006713796.1
7, 8
922
XM_011513081.2
XP_011511383.1
7, 8
921
XM_017007056.1
XP_016862545.1
3, 4
4447
XM_017007057.1
XP_016862546.1
4, 5
486
XM_017007058.1
XP_016862547.1
5, 6
599
XM_017007059.1
XP_016862548.1
3, 4
971
XM_017007060.1
XP_016862549.1
3, 4
561
XM_017007061.1
XP_016862550.1
3, 4
571
GenBank mRNA
AK027607.1
-
3, 4
371
AK123060.1
-
4, 5
1351
AK294979.1
-
7, 8
770
AK295091.1
-
6, 7
765
AK307847.1
-
6, 7
615
AL833312.1
-
4, 5
1490
BC040285.1
-
6, 7
726
4825732
BC042613.1
-
3, 4
3389
4822585
BX537652.1
-
3, 4
4822
DA031063.1
-
3, 4
442
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
substantia nigra development
protein localization to centrosome
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0